Merck
  • Effect of particle deposition parameters on silica coating of zirconia using a chairside air-abrasion device.
Effect of particle deposition parameters on silica coating of zirconia using a chairside air-abrasion device.
The journal of adhesive dentistry (2013-05-24)
Mutlu Ozcan, John Raadschelders, Pekka Vallittu, Lippo Lassilla
ABSTRACT
To evaluate the effect of nozzle distance, nozzle angle, and deposition duration on the silica content attained on zirconia by air abrasion. Disk-shaped zirconia (LAVA, 3M ESPE) (diameter: 10 mm, thickness: 2 mm) specimens (N = 54) were obtained. They were wet-ground finished using 600-, 800-, and 1200-grit silicone carbide abrasive papers in sequence and ultrasonically cleaned. The specimens were mounted in a specially designed apparatus that allowed the chairside air-abrasion device to be operated under standard conditions. Alumina-coated silica particles (CoJet Sand, 3M ESPE) were deposited on the zirconia disk surfaces varying the following parameters: a) nozzle distance (2, 5, 10 mm), and b) deposition duration (5, 13, 20 s) at two nozzle angles (45 and 90 degrees) under 2.5 bar pressure at three locations on each ceramic disk. The specimen surfaces were then gently air dried for 20 s. Silica content in weight percentage (wt%) was measured from 3 surfaces on each disk using Energy Dispersive X-ray Spectroscopy (EDS) (150X) in an area of 0.8 mm x 0.6 mm (n = 3 per group). Surface topographies were evaluated using SEM. Data were analyzed using ANOVA and Tukey's tests (α = 0.05). Nozzle angle (p = 0.003) and deposition duration (p = 0.03) significantly affected the results, but nozzle distance (p = 0.569) did not. A significantly higher amount of silica (wt%) was achieved when the nozzle angle was 45 degrees to the surface in all distance-duration combinations (16.7 to 28.2 wt%) compared to the 90-degree nozzle angle (10.7 to 18.6 wt%) (p < 0.001). The silica amount was significantly higher after 20-s deposition duration than after 13 s (p < 0.05). EDS analysis demonstrated not only Si but also Al, Zr, and O traces on the substrate. SEM images indicated that deposition at a nozzle distance of 2 mm often created cavitations in zirconia. Effective silica deposition using a charside air-abrasion device can be achieved when the nozzle is held at 45 degrees to the surface with more than 2-mm nozzle distance for 20 s.
Sigma-Aldrich
Silicon carbide, nanopowder, <100 nm particle size
Sigma-Aldrich
Aluminum, powder, <5 μm particle size, 99.5% trace metals basis
Sigma-Aldrich
Aluminum, foil, thickness 8 μm, 99% trace metals basis
Sigma-Aldrich
Zirconium, foil, thickness 0.1 mm, 99.98% trace metals basis
Sigma-Aldrich
Aluminum oxide, mesoporous, MSU-X (wormhole), average pore size 3.8 nm
Sigma-Aldrich
Aluminum, pellets, 3-8 mesh, ≥99.999% trace metals basis
Sigma-Aldrich
Aluminum, evaporation slug, diam. × L 6.3 mm × 6.3 mm, 99.999% trace metals basis
Sigma-Aldrich
Aluminum, wire, diam. 1.0 mm, 99.999% trace metals basis
Sigma-Aldrich
Aluminum, granular, <1 mm, 99.7% trace metals basis
Sigma-Aldrich
Aluminum, ≤30 μm, spherical powder, ≥99%
Sigma-Aldrich
Aluminum, powder, ≥91% (complexometric)
Aluminum, IRMM®, certified reference material, 1.0 mm foil
Sigma-Aldrich
Zirconium, rod, diam. 6.35 mm, ≥99% trace metals basis
Sigma-Aldrich
Silicon carbide, -200 mesh particle size
Sigma-Aldrich
Zirconium, sponge, ≥99% trace metals basis
Sigma-Aldrich
Zirconium, powder, −100 mesh
Sigma-Aldrich
Aluminum, powder, <75 μm, ≥99.95% trace metals basis
Sigma-Aldrich
Aluminum, wire, diam. 0.58 mm, ≥99.99% trace metals basis
Aluminum, IRMM®, certified reference material, 1.0 mm wire
Supelco
Aluminum oxide, activated, neutral, Brockmann Activity I
Sigma-Aldrich
Aluminum, ACS reagent, 99%, wire, wire diam. ~1.5 mm
Sigma-Aldrich
Silicon carbide, −400 mesh particle size, ≥97.5%
Sigma-Aldrich
Aluminum, ReagentPlus®, beads, 5-15 mm, 99.9% trace metals basis
Sigma-Aldrich
Aluminum oxide, 99.997% trace metals basis
Sigma-Aldrich
Zirconium(IV) oxide, 99.99% trace metals basis (purity excludes ~2% HfO2)
Supelco
Aluminum oxide, for the determination of hydrocarbons
Aluminum, IRMM®, certified reference material, 0.1 mm foil
Sigma-Aldrich
Silicon, pieces, 99.95% trace metals basis
Sigma-Aldrich
Zirconium(IV) oxide, nanopowder, <100 nm particle size (TEM)
Sigma-Aldrich
Aluminum, foil, thickness 0.25 mm, 99.999% trace metals basis