Merck
Mass spectrometry-based metabolomics of single yeast cells.
Proceedings of the National Academy of Sciences of the United States of America (2013-05-15)
Alfredo J Ibáñez, Stephan R Fagerer, Anna Mareike Schmidt, Pawel L Urban, Konstantins Jefimovs, Philipp Geiger, Reinhard Dechant, Matthias Heinemann, Renato Zenobi
ABSTRACT
Single-cell level measurements are necessary to characterize the intrinsic biological variability in a population of cells. In this study, we demonstrate that, with the microarrays for mass spectrometry platform, we are able to observe this variability. We monitor environmentally (2-deoxy-D-glucose) and genetically (ΔPFK2) perturbed Saccharomyces cerevisiae cells at the single-cell, few-cell, and population levels. Correlation plots between metabolites from the glycolytic pathway, as well as with the observed ATP/ADP ratio as a measure of cellular energy charge, give biological insight that is not accessible from population-level metabolomic data.
Sigma-Aldrich
2-Deoxy-D-glucose, ≥99% (GC), crystalline
Pricing and availability is not currently available.
Sigma-Aldrich
2-Deoxy-D-glucose, ≥98% (GC), BioXtra
Pricing and availability is not currently available.
Sigma-Aldrich
2-Deoxy-D-glucose, ≥98% (GC), crystalline
Pricing and availability is not currently available.