Merck
Assessing aluminium toxicity in streams affected by acid mine drainage.
Water science and technology : a journal of the International Association on Water Pollution Research (2013-04-13)
A S Waters, J G Webster-Brown
ABSTRACT
Acid mine drainage (AMD) has degraded water quality and ecology in streams on the Stockton Plateau, the site of New Zealand's largest open-cast coal mining operation. This has previously been attributed largely to the effects of acidity and elevated aluminium (Al) concentrations. However, the toxicity of dissolved Al is dependent on speciation, which is influenced by pH which affects Al hydrolysis, as well as the concentrations of organic carbon and sulphate which complex Al. Methods for the assessment of the toxic fraction of Al, by chemical analysis and geochemical modelling, have been investigated in selected streams on the Stockton Plateau, where dissolved Al concentrations ranged from 0.034 to 27 mg L(-1). Modelling using PHREEQC indicated that between 0.2 and 85% of the dissolved Al was present as the free ion Al(3+), the most toxic Al species, which dominated in waters of pH = 3.8-4.8. Al-sulphate complexation reduced the Al(3+) concentration at lower pH, while Al-organic and -hydroxide complexes dominated at higher pH. Macroinvertebrate richness in the streams identified an Al(3+) 'threshold' of approximately 0.42 mg/L, above which taxa declined rapidly. Colorimetric 'Aluminon' analysis on unpreserved, unfiltered waters provided a better estimation of Al(3+) concentrations than inductively couple plasma-mass spectrometry (ICP-MS) on filtered, acidified waters. The Aluminon method does not react with particulate Al or strong Al complexes, often registering as little as 53% of the dissolved Al concentration determined by ICP-MS.
Product Description
Aluminum, IRMM®, certified reference material, 1.0 mm foil
Aluminum, IRMM®, certified reference material, 1.0 mm wire
Sigma-Aldrich
Aluminum, pellets, 3-8 mesh, ≥99.999% trace metals basis
Sigma-Aldrich
Aluminum, evaporation slug, diam. × L 6.3 mm × 6.3 mm, 99.999% trace metals basis
Sigma-Aldrich
Aluminum, wire, diam. 1.0 mm, 99.999% trace metals basis
Sigma-Aldrich
Aluminum, granular, <1 mm, 99.7% trace metals basis
Sigma-Aldrich
Aluminum, ≤30 μm, spherical powder, ≥99%
Sigma-Aldrich
Aluminum, powder, <75 μm, ≥99.95% trace metals basis
Sigma-Aldrich
Aluminum, wire, diam. 0.58 mm, ≥99.99% trace metals basis
Sigma-Aldrich
Aluminum, powder, <5 μm particle size, 99.5% trace metals basis
Sigma-Aldrich
Aluminum, ACS reagent, 99%, wire, wire diam. ~1.5 mm
Sigma-Aldrich
Aluminum, powder, ≥91% (complexometric)
Sigma-Aldrich
Aluminum, foil, thickness 8 μm, 99% trace metals basis
Sigma-Aldrich
Aluminum, ReagentPlus®, beads, 5-15 mm, 99.9% trace metals basis
Sigma-Aldrich
Aluminum, CP
Sigma-Aldrich
Aluminum, foil, thickness 0.25 mm, 99.999% trace metals basis
Sigma-Aldrich
Aluminum, foil, thickness 0.13 mm, ≥99.99% trace metals basis
Sigma-Aldrich
Aluminum, foil, thickness 1.0 mm, 99.999% trace metals basis
Sigma-Aldrich
Aluminum, foil, thickness 0.45-0.55 mm, 99.999% trace metals basis
Sigma-Aldrich
Aluminum, pellets, 3-12 mm, 99.99% trace metals basis
Aluminum, IRMM®, certified reference material, 0.1 mm foil