Merck
  • The impact of aluminium smelter shut-down on the concentration of fluoride in vegetation and soils.
The impact of aluminium smelter shut-down on the concentration of fluoride in vegetation and soils.
Environmental pollution (Barking, Essex : 1987) (2013-04-03)
Kate M Brougham, Stephen R Roberts, Alan W Davison, Gordon R Port
ABSTRACT
Although a great deal is known about the deposition of fluoride on vegetation, and the hazards associated with uptake by grazing herbivores, little is known about what happens to the concentration of fluoride in vegetation and soil at polluted sites once deposition ceases. The closure of Anglesey Aluminium Metals Ltd smelter, in September 2009, provided a unique opportunity to study fluoride loading once deposition stopped. Fluoride was monitored in plants and soil within 1 km of the former emission source. Fluoride concentrations in a range of plant material had decreased to background levels of 10 mg F kg(-1) after 36 weeks. Concentrations of fluoride in mineral-rich soils decreased steadily demonstrating their limited potential to act as contaminating sources of fluoride for forage uptake. There were significant differences in the rate of decline of fluoride concentrations between plant species.
Sigma-Aldrich
Aluminum, CP
Sigma-Aldrich
Aluminum, foil, thickness 0.13 mm, ≥99.99% trace metals basis
Sigma-Aldrich
Aluminum, pellets, 3-12 mm, 99.99% trace metals basis
Sigma-Aldrich
Aluminum, foil, thickness 0.25 mm, 99.999% trace metals basis
Sigma-Aldrich
Aluminum, foil, thickness 1.0 mm, 99.999% trace metals basis
Aluminum, IRMM®, certified reference material, 0.1 mm foil
Sigma-Aldrich
Aluminum, foil, thickness 0.45-0.55 mm, 99.999% trace metals basis
Sigma-Aldrich
Aluminum, powder, <5 μm particle size, 99.5% trace metals basis
Sigma-Aldrich
Aluminum, foil, thickness 8 μm, 99% trace metals basis
Aluminum, IRMM®, certified reference material, 1.0 mm wire
Aluminum, IRMM®, certified reference material, 1.0 mm foil
Sigma-Aldrich
Aluminum, granular, <1 mm, 99.7% trace metals basis
Sigma-Aldrich
Aluminum, evaporation slug, diam. × L 6.3 mm × 6.3 mm, 99.999% trace metals basis
Sigma-Aldrich
Aluminum, ≤30 μm, spherical powder, ≥99%
Sigma-Aldrich
Aluminum, pellets, 3-8 mesh, ≥99.999% trace metals basis
Sigma-Aldrich
Aluminum, powder, <75 μm, ≥99.95% trace metals basis
Sigma-Aldrich
Aluminum, wire, diam. 1.0 mm, 99.999% trace metals basis
Sigma-Aldrich
Aluminum, ACS reagent, 99%, wire, wire diam. ~1.5 mm
Sigma-Aldrich
Aluminum, powder, ≥91% (complexometric)
Sigma-Aldrich
Aluminum, wire, diam. 0.58 mm, ≥99.99% trace metals basis
Sigma-Aldrich
Aluminum, ReagentPlus®, beads, 5-15 mm, 99.9% trace metals basis