Merck
  • Synthesis and self-assembly of photonic materials from nanocrystalline titania sheets.
Synthesis and self-assembly of photonic materials from nanocrystalline titania sheets.
Journal of the American Chemical Society (2013-03-19)
Jian Zhu, Jinguo Wang, Fujian Lv, Shengxiong Xiao, Colin Nuckolls, Hexing Li
ABSTRACT
We describe the use of benzyl alcohols in a solvothermal/alcoholysis reaction to form nanocrystalline sheets of anatase titania. By tuning the reaction conditions, we adjust the size of the nanosheets. The type and density of benzyl groups that decorate the basal plane of the titania sheets control the self-assembly into layered structures. These layered materials can be grown from solid substrates to create iridescent thin films that reflect specific wavelengths of visible light.
Sigma-Aldrich
Benzyl alcohol, SAJ special grade, ≥99.0%
Sigma-Aldrich
Benzyl alcohol, SAJ first grade, ≥98.5%
Sigma-Aldrich
Benzyl alcohol, ≥99%, FCC, FG
Sigma-Aldrich
Benzyl alcohol, natural, ≥98%, FG
Sigma-Aldrich
Benzyl alcohol, anhydrous, 99.8%
Supelco
Benzyl alcohol, analytical standard
Supelco
Benzyl alcohol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Benzyl alcohol, puriss. p.a., ACS reagent, ≥99.0% (GC)
Sigma-Aldrich
Benzyl alcohol, ReagentPlus®, ≥99%
Sigma-Aldrich
Benzyl alcohol, ACS reagent, ≥99.0%
Sigma-Aldrich
Benzyl alcohol, puriss., meets analytical specification of Ph. Eur., BP, NF, 99-100.5% (GC)
Benzyl alcohol, European Pharmacopoeia (EP) Reference Standard