Merck
  • Peroxisomal catalase deficiency modulates yeast lifespan depending on growth conditions.
Peroxisomal catalase deficiency modulates yeast lifespan depending on growth conditions.
Aging (2013-02-22)
Adam Kawałek, Sophie D Lefevre, Marten Veenhuis, Ida J van der Klei
ABSTRACT
We studied the role of peroxisomal catalase in chronological aging of the yeastHansenula polymorpha in relation to various growth substrates. Catalase-deficient (cat) cells showed a similar chronological life span (CLS) relative to the wild-type control upon growth on carbon and nitrogen sources that are not oxidized by peroxisomal enzymes. However, when media contained methylamine, which is oxidized by peroxisomal amine oxidase, the CLS of cat cells was significantly reduced. Conversely, the CLS of cat cells was enhanced relative to the wild-type control, when cells were grown on methanol, which is oxidized by peroxisomal alcohol oxidase. At these conditions strongly enhanced ROS levels were observed during the exponential growth phase of cat cells. This was paralleled by activation of the transcription factor Yap1, as well as an increase in the levels of the antioxidant enzymes cytochrome c peroxidase and superoxide dismutase. Upon deletion of the genes encoding Yap1 or cytochrome c peroxidase, the CLS extension of cat cells on methanol was abolished. These findings reveal for the first time an important role of enhanced cytochrome c peroxidase levels in yeast CLS extension.
Sigma-Aldrich
Nonne-Apelt′s reagent
Pricing and availability is not currently available.
Sigma-Aldrich
Ammonium-14N2 sulfate solution, 40 wt. % in H2O, 99.99 atom % 14N
Pricing and availability is not currently available.
Sigma-Aldrich
Methylamine solution, 40 wt. % in H2O
Pricing and availability is not currently available.
Sigma-Aldrich
Ammonium sulfate-14N2 solution, 40 wt. % in H2O, 99.99 atom % 14N
Pricing and availability is not currently available.
Sigma-Aldrich
Methylamine-13C hydrochloride, 99 atom % 13C
Pricing and availability is not currently available.
Sigma-Aldrich
Methylamine solution, 33 wt. % in absolute ethanol ((denatured with 1% toluene))
Pricing and availability is not currently available.
Sigma-Aldrich
Methylamine solution, 2.0 M in methanol
Pricing and availability is not currently available.
Sigma-Aldrich
Methylamine solution, 2.0 M in THF
Pricing and availability is not currently available.
Sigma-Aldrich
Ammonium sulfate, SAJ first grade, ≥99.0%
Pricing and availability is not currently available.
Supelco
Ammonium sulfate, analytical standard, for Nitrogen Determination According to Kjeldahl Method, traceable to NIST SRM 194
Pricing and availability is not currently available.
Sigma-Aldrich
Ammonium sulfate, 99.999% trace metals basis
Pricing and availability is not currently available.
Sigma-Aldrich
Ammonium sulfate, suitable for plant cell culture, ≥99.0%
Pricing and availability is not currently available.
Sigma-Aldrich
Ammonium sulfate, BioXtra, ≥99.0%
Pricing and availability is not currently available.
Sigma-Aldrich
Methylamine hydrochloride, ≥98%
Pricing and availability is not currently available.
Sigma-Aldrich
Ammonium sulfate, for molecular biology, ≥99.0%
Pricing and availability is not currently available.
Sigma-Aldrich
Ammonium sulfate, BioUltra, ≥99.0% (T)
Pricing and availability is not currently available.
Sigma-Aldrich
Methylamine-15N hydrochloride, 98 atom % 15N
Pricing and availability is not currently available.
Sigma-Aldrich
Ammonium sulfate, ≥99.5%
Pricing and availability is not currently available.
Sigma-Aldrich
Ammonium sulfate, anhydrous, Redi-Dri, ReagentPlus®, ≥99.0%
Pricing and availability is not currently available.
Sigma-Aldrich
Ammonium sulfate, ACS reagent, ≥99.0%
Pricing and availability is not currently available.
Sigma-Aldrich
Ammonium sulfate, ReagentPlus®, ≥99.0%
Pricing and availability is not currently available.
Sigma-Aldrich
Ammonium sulfate, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99.0%
Pricing and availability is not currently available.