Merck
  • Biochemical similarities and differences between the catalytic [4Fe-4S] cluster containing fumarases FumA and FumB from Escherichia coli.
Biochemical similarities and differences between the catalytic [4Fe-4S] cluster containing fumarases FumA and FumB from Escherichia coli.
PloS one (2013-02-14)
Barbara M A van Vugt-Lussenburg, Laura van der Weel, Wilfred R Hagen, Peter-Leon Hagedoorn
ABSTRACT
The highly homologous [4Fe-4S] containing fumarases FumA and FumB, sharing 90% amino acid sequence identity, from Escherichia coli are differentially regulated, which suggests a difference in their physiological function. The ratio of FumB over FumA expression levels increases by one to two orders of magnitude upon change from aerobic to anaerobic growth conditions. To understand this difference in terms of structure-function relations, catalytic and thermodynamic properties were determined for the two enzymes obtained from homologous overexpression systems. FumA and FumB are essentially identical in their Michaelis-Menten kinetics of the reversible fumarate to L-malate conversion; however, FumB has a significantly greater catalytic efficiency for the conversion of D-tartrate to oxaloacetate consistent with the requirement of the fumB gene for growth on D-tartrate. Reduction potentials of the [4Fe-4S](2+) Lewis acid active centre were determined in mediated bulk titrations in the presence of added substrate and were found to be approximately -290 mV for both FumA and FumB. This study contradicts previously published claims that FumA and FumB exhibit different catalytic preferences for the natural substrates L-malate and fumarate. FumA and FumB differ significantly only in the catalytic efficiency for the conversion of D-tartrate, a supposedly non-natural substrate. The reduction potential of the substrate-bound [4Fe-4S] active centre is, contrary to previously reported values, close to the cellular redox potential.
Sigma-Aldrich
DL-Tartaric acid, ≥99%
Sigma-Aldrich
Fumarase from porcine heart, ammonium sulfate suspension, ≥300 units/mg protein (biuret)
Sigma-Aldrich
Oxaloacetic acid, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥97% (HPLC)
Sigma-Aldrich
Oxaloacetic acid, powder, suitable for hybridoma
Sigma-Aldrich
Oxaloacetic acid, ≥97% (HPLC)
Sigma-Aldrich
DL-Tartaric acid, ReagentPlus®, 99%
Sigma-Aldrich
L-(+)-Tartaric acid, JIS special grade, ≥99.5%
Sigma-Aldrich
L-(+)-Tartaric acid, SAJ first grade, ≥99.5%
Supelco
L-(+)-Tartaric acid, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
D-(−)-Tartaric acid, puriss., unnatural form, ≥99.0% (T)
Sigma-Aldrich
L-(+)-Tartaric acid, ≥99.7%, FCC, FG
Sigma-Aldrich
D-(−)-Tartaric acid, ReagentPlus®, 99%
Sigma-Aldrich
L-(+)-Tartaric acid, ≥99.5%
Sigma-Aldrich
L-(+)-Tartaric acid, ACS reagent, ≥99.5%
Sigma-Aldrich
L-(+)-Tartaric acid, puriss. p.a., reag. ISO, 99.5-101.0% (calc. to the dried substance)
Sigma-Aldrich
L-(+)-Tartaric acid, BioXtra
Sigma-Aldrich
L-(+)-Tartaric acid, puriss., meets analytical specification of Ph. Eur., NF, 99.7-100.5% (calc. to the dried substance), powder
Sigma-Aldrich
L-(+)-Tartaric acid, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99.5%