Merck
  • Chromogenic and fluorogenic chemosensors and reagents for anions. A comprehensive review of the years 2010-2011.
Chromogenic and fluorogenic chemosensors and reagents for anions. A comprehensive review of the years 2010-2011.
Chemical Society reviews (2013-02-13)
Luis E Santos-Figueroa, María E Moragues, Estela Climent, Alessandro Agostini, Ramón Martínez-Máñez, Félix Sancenón
ABSTRACT
This review focuses on examples reported in the years 2010-2011 dealing with the design of chromogenic and fluorogenic chemosensors or reagents for anions.
Product Description
Supelco
Benzyl alcohol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Benzyl alcohol, SAJ special grade, ≥99.0%
Sigma-Aldrich
Benzyl alcohol, SAJ first grade, ≥98.5%
Benzyl alcohol, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Benzyl alcohol, anhydrous, 99.8%
Supelco
Benzyl alcohol, analytical standard
Sigma-Aldrich
Benzyl alcohol, ≥99%, FCC, FG
Sigma-Aldrich
Benzyl alcohol, natural, ≥98%, FG
Sigma-Aldrich
Benzyl alcohol, puriss. p.a., ACS reagent, ≥99.0% (GC)
Sigma-Aldrich
Benzyl alcohol, ReagentPlus®, ≥99%
Sigma-Aldrich
Benzyl alcohol, ACS reagent, ≥99.0%
Sigma-Aldrich
Benzyl alcohol, puriss., meets analytical specification of Ph. Eur., BP, NF, 99-100.5% (GC)
Supelco
Benzyl alcohol, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland