Merck
  • Does correction of metabolic acidosis slow chronic kidney disease progression?
Does correction of metabolic acidosis slow chronic kidney disease progression?
Current opinion in nephrology and hypertension (2013-02-06)
Nimrit Goraya, Donald E Wesson
ABSTRACT
Most patients with chronic kidney disease (CKD) have progressive decline in glomerular filtration rate (GFR), despite current treatment practices. Recent studies support that dietary acid reduction with oral sodium based alkali or base-inducing food types add kidney protection to that provided by current kidney-protective interventions. Related studies also support that correction of metabolic acidosis with dietary acid reduction slows CKD progression. We reviewed these recent studies that show improvement in CKD parameters and slower CKD progression in response to improvement of CKD-associated metabolic acidosis with these interventions. Animal as well as human models of CKD show that alkali treatment ameliorates indices of kidney injury and also might slow GFR decline in patients with or without metabolic acidosis. These benefits have been similar with oral sodium-based alkali and base-inducing fruits and vegetables, supporting dietary acid reduction as an effective adjunct to conventional kidney-protective interventions. Recent studies suggest that metabolic acidosis mediates nephropathy progression, and its treatment with the comparatively inexpensive and well tolerated intervention of dietary acid reduction holds promise to be an additional kidney-protective strategy in CKD management.
Product Description
Supelco
Sodium bicarbonate concentrate, 0.1 M NaHCO3 in water, eluent concentrate for IC
Sigma-Aldrich
Sodium bicarbonate, tested according to Ph. Eur.
Sigma-Aldrich
Sodium bicarbonate, powder, BioReagent, for molecular biology, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Sodium bicarbonate solution, solution (7.5%), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Sodium bicarbonate, BioXtra, 99.5-100.5%
Sigma-Aldrich
Sodium bicarbonate, ReagentPlus®, ≥99.5%, powder
Sigma-Aldrich
Sodium bicarbonate, ACS reagent, ≥99.7%
Sigma-Aldrich
Sodium bicarbonate, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Sodium bicarbonate, puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, E500, 99.0-100.5%, powder
Sigma-Aldrich
Sodium hydrogencarbonate, −40-+140 mesh, ≥95%
Sigma-Aldrich
Sodium bicarbonate-12C, 99.9 atom % 12C