Merck
  • A rechargeable Li-O2 battery using a lithium nitrate/N,N-dimethylacetamide electrolyte.
A rechargeable Li-O2 battery using a lithium nitrate/N,N-dimethylacetamide electrolyte.
Journal of the American Chemical Society (2013-01-31)
Wesley Walker, Vincent Giordani, Jasim Uddin, Vyacheslav S Bryantsev, Gregory V Chase, Dan Addison
ABSTRACT
A major challenge in the development of rechargeable Li-O(2) batteries is the identification of electrolyte materials that are stable in the operating environment of the O(2) electrode. Straight-chain alkyl amides are one of the few classes of polar, aprotic solvents that resist chemical degradation in the O(2) electrode, but these solvents do not form a stable solid-electrolyte interphase (SEI) on the Li anode. The lack of a persistent SEI leads to rapid and sustained solvent decomposition in the presence of Li metal. In this work, we demonstrate for the first time successful cycling of a Li anode in the presence of the solvent, N,N-dimethylacetamide (DMA), by employing a salt, lithium nitrate (LiNO(3)), that stabilizes the SEI. A Li-O(2) cell containing this electrolyte composition is shown to cycle for more than 2000 h (>80 cycles) at a current density of 0.1 mA/cm(2) with a consistent charging profile, good capacity retention, and O(2) detected as the primary gaseous product formed during charging. The discovery of an electrolyte system that is compatible with both electrodes in a Li-O(2) cell may eliminate the need for protecting the anode with a ceramic membrane.
Sigma-Aldrich
Lithium nitrate, ReagentPlus®
Sigma-Aldrich
N,N-Dimethylacetamide, SAJ first grade, ≥99.0%
Sigma-Aldrich
N,N-Dimethylacetamide, SAJ special grade
Supelco
N,N-Dimethylacetamide, analytical standard
Sigma-Aldrich
N,N-Dimethylacetamide, ReagentPlus®, ≥99%
Sigma-Aldrich
N,N-Dimethylacetamide, anhydrous, 99.8%
Sigma-Aldrich
N,N-Dimethylacetamide, suitable for peptide synthesis, ≥99.8% (GC)
Sigma-Aldrich
N,N-Dimethylacetamide, spectrophotometric grade, ≥99%
Sigma-Aldrich
N,N-Dimethylacetamide, puriss. p.a., ≥99.5% (GC)
Sigma-Aldrich
N,N-Dimethylacetamide, ReagentPlus®, 99%
Sigma-Aldrich
N,N-Dimethylacetamide, suitable for HPLC, ≥99.9%