Merck
  • Surface-modified anodic aluminum oxide membrane with hydroxyethyl celluloses as a matrix for bilirubin removal.
Surface-modified anodic aluminum oxide membrane with hydroxyethyl celluloses as a matrix for bilirubin removal.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences (2013-01-08)
Maoqiang Xue, Yisheng Ling, Guisen Wu, Xin Liu, Dongtao Ge, Wei Shi
ABSTRACT
Microporous anodic aluminum oxide (AAO) membranes were modified by 3-glycidoxypropyltrimethoxysilane to produce terminal epoxy groups. These were used to covalently link hydroxyethyl celluloses (HEC) to amplify reactive groups of AAO membrane. The hydroxyl groups of HEC-AAO composite membrane were further modified with 1,4-butanediol diglycidyl ether to link arginine as an affinity ligand. The contents of HEC and arginine of arginine-immobilized HEC-AAO membrane were 52.1 and 19.7mg/g membrane, respectively. As biomedical adsorbents, the arginine-immobilized HEC-AAO membranes were tested for bilirubin removal. The non-specific bilirubin adsorption on the unmodified HEC-AAO composite membranes was 0.8mg/g membrane. Higher bilirubin adsorption values, up to 52.6mg/g membrane, were obtained with the arginine-immobilized HEC-AAO membranes. Elution of bilirubin showed desorption ratio was up to 85% using 0.3M NaSCN solution as the desorption agent. Comparisons equilibrium and dynamic capacities showed that dynamic capacities were lower than the equilibrium capacities. In addition, the adsorption mechanism of bilirubin and the effects of temperature, initial concentration of bilirubin, albumin concentration and ionic strength on adsorption were also investigated.
Sigma-Aldrich
Hydroxyethyl-cellulose
Sigma-Aldrich
Hydroxyethyl-cellulose, viscosity 90-160 cP, 5 % in H2O(25 °C)
Sigma-Aldrich
2-Hydroxyethyl cellulose, average Mv ~1,300,000
Sigma-Aldrich
2-Hydroxyethyl cellulose, average Mv ~720,000
Sigma-Aldrich
2-Hydroxyethyl cellulose, average Mw ~380,000
Sigma-Aldrich
2-Hydroxyethyl cellulose, average Mv ~90,000