Merck
Inhibition of human estrogen synthetase (aromatase) by flavones.
Science (New York, N.Y.) (1984-09-07)
J T Kellis, L E Vickery
ABSTRACT
Several naturally occurring and synthetic flavones were found to inhibit the aromatization of androstenedione and testosterone to estrogens catalyzed by human placental and ovarian microsomes. These flavones include (in order of decreasing potency) 7,8-benzoflavone, chrysin, apigenin, flavone, flavanone, and quercetin; 5,6-benzoflavone was not inhibitory. 7,8-Benzoflavone and chrysin were potent competitive inhibitors and induced spectral changes in the aromatase cytochrome P-450 indicative of substrate displacement. Flavones may thus compete with steroids in their interaction with certain monooxygenases and thereby alter steroid hormone metabolism.
Sigma-Aldrich
Apigenin, ≥95.0% (HPLC)