Merck
  • Dextromethorphan attenuated inflammation and combined opioid use in humans undergoing methadone maintenance treatment.
Dextromethorphan attenuated inflammation and combined opioid use in humans undergoing methadone maintenance treatment.
Journal of neuroimmune pharmacology : the official journal of the Society on NeuroImmune Pharmacology (2012-09-20)
Shiou-Lan Chen, Sheng-Yu Lee, Pao-Luh Tao, Yun-Hsuan Chang, Shih-Heng Chen, Chun-Hsien Chu, Po See Chen, I Hui Lee, Tzung Lieh Yeh, Yen Kuang Yang, Jau-Shyong Hong, Ru-Band Lu
ABSTRACT
Recent studies show that proinflammatory cytokines might be related to the development of opioid dependence (physiological, psychological, or both). In a double-blind, randomly stratified clinical trial investigating whether add-on dextromethorphan (60-120 mg/day) attenuated inflammation and the combined use of opioids in heroin-dependent patients undergoing methadone maintenance treatment, we evaluated whether inflammation is related to the progression of opioid dependence. All participants (107 heroin-dependent patients and 84 nondependent healthy controls) were recruited from National Cheng Kung University Hospital. Their plasma cytokine levels were measured to evaluate the effect of add-on dextromethorphan. Plasma TNF-α and IL-8 levels were significantly higher in long-term heroin-dependent patients than in healthy controls (p < 0.001). Chronic heroin-use-induced TNF-α and IL-8 levels were significantly (p < 0.05) attenuated in patients treated for 12 weeks with add-on dextromethorphan. Moreover, both tolerance to methadone and the combined use of opioids were significantly (p < 0.05) attenuated in patients taking dextromethorphan. We conclude that dextromethorphan might be a feasible adjuvant therapeutic for attenuating inflammation and inhibiting methadone tolerance and combined opioid use in heroin-dependent patients.
Sigma-Aldrich
Dextromethorphan hydrobromide monohydrate, ≥99% (TLC)
Sigma-Aldrich
Dextromethorphan hydrobromide, meets USP testing specifications
Supelco
Dextromethorphan HBr, certified reference material, pharmaceutical secondary standard
Dextromethorphan hydrobromide, European Pharmacopoeia (EP) Reference Standard
Supelco
Dextromethorphan solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®