Merck
  • AlCl3 mediated unexpected migration of sulfonyl groups: regioselective synthesis of 7-sulfonyl indoles of potential pharmacological interest.
AlCl3 mediated unexpected migration of sulfonyl groups: regioselective synthesis of 7-sulfonyl indoles of potential pharmacological interest.
Chemical communications (Cambridge, England) (2012-09-20)
Bagineni Prasad, Raju Adepu, Sandhya Sandra, D Rambabu, G Rama Krishna, C Malla Reddy, Girdhar Singh Deora, Parimal Misra, Manojit Pal
ABSTRACT
A conceptually new and straightforward introduction of sulfonyl groups at the C-7 position of an indole ring has been achieved via AlCl(3) mediated unexpected regioselective sulfonyl group migration for N-alkyl/aryl/heteroarylsulfonyl indoles affording potential inhibitors of Mycobacterium tuberculosis H37Rv chorismate mutase.
Sigma-Aldrich
Aluminum chloride, anhydrous, powder, 99.99% trace metals basis
Sigma-Aldrich
Aluminum chloride, 99.99% trace metals basis
Sigma-Aldrich
Aluminum chloride, anhydrous, powder, 99.999% trace metals basis
Sigma-Aldrich
Aluminum chloride, SAJ first grade, ≥98.0%
Sigma-Aldrich
Aluminum chloride solution, 1.0 M in nitrobenzene
Sigma-Aldrich
Aluminum chloride, ReagentPlus®, 99%
Sigma-Aldrich
Aluminum chloride, anhydrous, sublimed, ≥98%
Sigma-Aldrich
Aluminum chloride, reagent grade, 98%
Sigma-Aldrich
Aluminum chloride hydrate, 99.999% trace metals basis