Merck
Photosensitizing properties of compounds related to benzophenone.
Acta dermato-venereologica (2012-09-18)
Marianne Placzek, Markus Dendorfer, Bernhard Przybilla, Klaus-P Gilbertz, Bernadette Eberlein
ABSTRACT
Benzophenone is a phototoxic compound with absorption maxima in the ultraviolet A (UVA) and ultraviolet B (UVB) range. Many benzophenone derivatives are known to be photosensitizing. On the other hand, 2-hydroxy-4-methoxybenzophenone is used as a photoprotective agent. The aim of the present study was to analyse a range of benzophenone derivatives and thus examine the effects of molecular changes in the benzophenone molecule on phototoxic behaviour. Phototoxicity was tested by an in vitro photohaemolysis test. The tested compounds were benzophenone itself and the derivatives 2-hydroxybenzophenone, 2-aminobenzophenone, 2-benzoylbenzoic acid, 3-hydroxybenzophenone, and 4-hydroxybenzo-phenone, as well as the structurally similar compounds 9-fluorenone, 9-fluorenone-2-carboxylic acid, cyclohexyl phenyl ketone, and 1,4-naphtho-quinone. It was shown that minor changes in molecular structure can result in highly different phototoxic characteristics.
Product Description
Supelco
Melting point standard 47-49°C, analytical standard
Sigma-Aldrich
2-Benzoylbenzoic acid, 98%
Supelco
Mettler-Toledo Calibration substance ME 18870, Benzophenone, traceable to primary standards (LGC)
Sigma-Aldrich
Benzophenone, SAJ first grade, ≥99.0%
Sigma-Aldrich
1,4-Naphthoquinone, purum, ≥96.5% (HPLC)
Sigma-Aldrich
4-Hydroxybenzophenone, 98%
Sigma-Aldrich
Cyclohexyl phenyl ketone, 98%
Sigma-Aldrich
Benzophenone, ReagentPlus®, 99%
Sigma-Aldrich
9-Fluorenone, 98%
Sigma-Aldrich
1,4-Naphthoquinone, 97%
Supelco
Benzophenone, analytical standard
Sigma-Aldrich
Benzophenone, purified by sublimation, ≥99%
Supelco
Benzophenone, Pharmaceutical Secondary Standard; Certified Reference Material
Benzophenone, European Pharmacopoeia (EP) Reference Standard