Merck
  • Ruthenium-catalyzed transformation of aryl and alkenyl triflates to halides.
Ruthenium-catalyzed transformation of aryl and alkenyl triflates to halides.
Journal of the American Chemical Society (2012-08-25)
Yusuke Imazaki, Eiji Shirakawa, Ryota Ueno, Tamio Hayashi
ABSTRACT
Aryl triflates were transformed to aryl bromides/iodides simply by treating them with LiBr/NaI and [Cp*Ru(MeCN)(3)]OTf. The ruthenium complex also catalyzed the transformation of alkenyl sulfonates and phosphates to alkenyl halides under mild conditions. Aryl and alkenyl triflates undergo oxidative addition to a ruthenium(II) complex to form η(1)-arylruthenium and 1-ruthenacyclopropene intermediates, respectively, which are transformed to the corresponding halides.
Sigma-Aldrich
Silver trifluoromethanesulfonate, ≥99%
Sigma-Aldrich
Aluminum trifluoromethanesulfonate, 99.9% trace metals basis
Sigma-Aldrich
Silver trifluoromethanesulfonate, purum, ≥98.0% (Ag)
Sigma-Aldrich
Silver trifluoromethanesulfonate, ≥99.95% trace metals basis
Sigma-Aldrich
Trifluoromethanesulfonic acid, reagent grade, 98%
Sigma-Aldrich
Zinc trifluoromethanesulfonate, 98%
Sigma-Aldrich
Lanthanum(III) trifluoromethanesulfonate, 99.999% trace metals basis
Sigma-Aldrich
Trifluoromethanesulfonic acid, ReagentPlus®, ≥99%
Sigma-Aldrich
Lithium trifluoromethanesulfonate, 96%
Sigma-Aldrich
Lithium trifluoromethanesulfonate, 99.995% trace metals basis