Merck
  • Reactivity of ruthenium vinylidene complexes containing indenyl/dppe ligands and unsaturated bonds at Cδ with trimethylsilyl azide.
Reactivity of ruthenium vinylidene complexes containing indenyl/dppe ligands and unsaturated bonds at Cδ with trimethylsilyl azide.
Molecules (Basel, Switzerland) (2012-07-19)
Hui-Ling Sung, Tze-Min Her, Wen-Hsien Su, Chin-Pao Cheng
ABSTRACT
This study presents a new reaction of cationic vinylidene complexes with Me₃SiN₃ (TMSN₃), which yields N-coordinated nitrile complexes 3. Treatment of a ruthenium acetylide precursor containing indenyl and dppe ligands with a series of organic halides produced the corresponding vinylidene complexes 2 in good yield. Further reaction of 2 with TMSN₃ at room temperature produced N-coordinated ruthenium nitrile complexes 3. Unlike the reaction of cyclopropenylruthenium complexes with TMSN₃, which yielded different products depending on the substituent at Cγ, the vinylidene complexes containing unsaturated bonds at Cd yielded similar N-coordinated nitrile complexes. This transformation did not seemingly occur in the reaction of ruthenium vinylidene complexes containing Cp and PPh₃ ligands with TMSN₃. Deprotonation of these vinylidene complexes yielded cyclopropenyl or thermodynamic furylruthenium complexes, depending on the substitute at Cγ. Subsequent reactions of the cyclopropenyl or furylruthenium complexes with TMSN₃ afforded different products.
Sigma-Aldrich
1,2-Dipalmitoyl-sn-glycero-3-phosphoethanolamine, ≥97%
Sigma-Aldrich
1,2-Dipalmitoyl-rac-glycero-3-phosphoethanolamine, synthetic, ≥98%