Merck
  • Enhancing starch production of a marine green microalga Tetraselmis subcordiformis through nutrient limitation.
Enhancing starch production of a marine green microalga Tetraselmis subcordiformis through nutrient limitation.
Bioresource technology (2012-06-22)
Changhong Yao, Jiangning Ai, Xupeng Cao, Song Xue, Wei Zhang
ABSTRACT
Microalgal starch is a potential feedstock for biofuel production. The effects of KNO(3) and MgSO(4) concentrations and light intensity on biomass and starch production by the marine microalga, Tetraselmis subcordiformis, were investigated. Under 200 μmol m(-2) s(-1) irradiance and sulfur-deprived conditions, a starch productivity of 0.62 g L(-1) d(-1) and a starch content of 62.1% based on dry weight (DW) was achieved. A starch content of 54.3% was achieved under low irradiance and nitrogen starvation, which was 6.5% higher than that under nutrient- and light-sufficient conditions. Photosynthetic activity was indispensable for starch accumulation. It is difficult to reach high starch productivity and starch concentration simultaneously. Proper nutrient concentrations are necessary to achieve high starch productivity or starch concentration based on the target. The high starch productivity and starch content suggest that T. subcordiformis is a promising microalgal starch producer.
Sigma-Aldrich
Potassium nitrate, 99.99% trace metals basis
Sigma-Aldrich
Potassium nitrate, BioXtra, ≥99.0%
Sigma-Aldrich
Potassium nitrate, BioReagent, suitable for cell culture, suitable for plant cell culture
Sigma-Aldrich
Potassium nitrate, SAJ first grade, ≥99.0%
Sigma-Aldrich
Potassium nitrate, 99.999% trace metals basis
Sigma-Aldrich
Potassium nitrate, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Potassium nitrate, ACS reagent, ≥99.0%
Sigma-Aldrich
Potassium nitrate, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99%
Sigma-Aldrich
Potassium nitrate, puriss., meets analytical specification of Ph Eur, BP, USP, FCC, E252, 99.0-100.5%
Supelco
Mettler-Toledo Calibration substance ME 51143095, Potassium nitrate, traceable to primary standards (LGC)