Merck
  • Oxidized dextran as crosslinker for chitosan cryogel scaffolds and formation of polyelectrolyte complexes between chitosan and gelatin.
Oxidized dextran as crosslinker for chitosan cryogel scaffolds and formation of polyelectrolyte complexes between chitosan and gelatin.
Macromolecular bioscience (2012-06-08)
Dmitriy Berillo, Linda Elowsson, Harald Kirsebom
ABSTRACT
Macroporous scaffolds composed of chitosan and using oxidized dextran as a crosslinker are produced through cryogelation. Introducing gelatin as a third component into the structure results in the formation of mesopores in the pore walls, which are not seen if gelatin is excluded. The mesoporous structure is explained by the formation of polyelectrolyte complexes between chitosan and gelatin before crosslinking takes place. The scaffolds exhibit highly elastic properties withstanding compressions up to 60%. The in vitro biocompatibility of the cryogels is evaluated using fibroblasts from a mouse cell line (L929) and it is seen that the cells adhere and proliferate on the scaffolds. The mesoporous structure seems to have a positive effect on proliferation.
Sigma-Aldrich
Sodium (meta)periodate, BioUltra, ≥99.5% (RT)
Sigma-Aldrich
Sodium (meta)periodate, ≥99.0%
Sigma-Aldrich
Periodic acid, 99.999% trace metals basis
Sigma-Aldrich
Periodic acid, suitable for electrophoresis, ≥99%
Sigma-Aldrich
Periodic acid, SAJ special grade, ≥98.5%
Sigma-Aldrich
Sodium periodate, ACS reagent, ≥99.8%
Sigma-Aldrich
Periodic acid, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Periodic acid, puriss. p.a., ACS reagent, for oxidimetric titration, crystallized, ≥99.0% (RT)
Sigma-Aldrich
Periodic acid, ACS reagent, 99%