Merck
  • Visualizing hydrophobic domains in silicone hydrogel lenses with Sudan IV.
Visualizing hydrophobic domains in silicone hydrogel lenses with Sudan IV.
Investigative ophthalmology & visual science (2012-04-19)
Jean T Jacob, Jacques Levet, Tamika A Edwards, Nissanke Dassanayake, Howard Ketelson
ABSTRACT
A lipophilic dye is used to investigate the degree to which the surface and bulk hydrophobic domains of the lenses can be imaged and to identify specific changes in the availability of those domains after in vitro wear and cleaning conditions. The effect of a multipurpose solution (MPS), OPTI-FREE RepleniSH, on lens hydrophobic domains was also investigated. Hydrophobic domains were determined using a saturated solution of Sudan IV. Staining periods of 30 minutes and 16 hours were used to determine surface versus bulk hydrophobic domains. Four types of silicone hydrogel lens materials were tested. The degree of staining was visually documented by photography and quantitatively determined by extraction and analysis of the total amount of dye adsorbed. Specific differences in staining were found for all control lenses. Exposure to in vitro wear conditions significantly decreased the staining response for all lens types as compared with unworn lenses (P = 0.001). However, the trend of staining remained the same: balafilcon A > galyfilcon A > senofilcon A > lotrafilcon B. MPS decreased the extent of staining; the degree of its effect varied with lens type. Hydrophobic staining with Sudan IV visualized domains on and within silicone hydrogel lenses. Differences in staining response after exposure to wear and cleaning conditions indicate the potential for protein and lipid deposition on the different lens types and the ability of MPS to affect that deposition. Hydrophobic staining may be useful for determining differences in surface modification and lipophilicity of silicone hydrogel lenses.
Sigma-Aldrich
Sudan IV, certified by the BSC, certified by the Biological Stain Commission, BioXtra
Supelco
Sudan IV, analytical standard
Sigma-Aldrich
Sudan IV, Certified Biological Stain Commission,Dye content ≥80 %