Merck
The effect of a mild base on curcumin in methanol and ethanol.
The journal of physical chemistry. A (2012-02-10)
Yuval Erez, Itay Presiado, Rinat Gepshtein, Dan Huppert
ABSTRACT
Steady-state and time-resolved emission techniques were employed to study the effect of acetate, a mild base, on the luminescence of curcumin in methanol and ethanol. We found that the steady-state emission intensity as well as the average fluorescence decay time are reduced by a factor of 5 when the acetate concentration is raised to about 1.8 M. We attribute this large effect to an excited-state proton transfer (ESPT) from the acidic groups of curcumin to the acetate anion. We analyze the experimental data in terms of an ESPT reaction occurring between a photoacid and a base.
Sigma-Aldrich
Potassium acetate solution, BioUltra, for molecular biology, 5 M in H2O
Sigma-Aldrich
Potassium acetate, SAJ first grade, ≥95.0%
Sigma-Aldrich
Potassium acetate, 99.98% trace metals basis
Sigma-Aldrich
Potassium acetate, BioUltra, for molecular biology, ≥99.0% (NT)
Sigma-Aldrich
Potassium acetate, for molecular biology, ≥99.0%
Sigma-Aldrich
Potassium acetate, ACS reagent, ≥99.0%
Sigma-Aldrich
Potassium acetate, BioXtra, ≥99.0%
Sigma-Aldrich
Potassium acetate, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Potassium acetate, meets USP testing specifications
Sigma-Aldrich
Potassium acetate, puriss., meets analytical specification of Ph. Eur., BP, E261, 99-101%