Merck
  • Automatized flow-batch method for fluorescent determination of free glycerol in biodiesel samples using on-line extraction.
Automatized flow-batch method for fluorescent determination of free glycerol in biodiesel samples using on-line extraction.
Talanta (2012-01-31)
Marcelo B Lima, Matías Insausti, Claudia E Domini, Marcelo F Pistonesi, Mário C Ugulino de Araújo, Beatriz S Fernández Band
ABSTRACT
An automatic method, based on flow-batch (FB), for determining glycerol in biodiesel was developed. The FB systems draw upon the useful features of flow, batch and multi-commutation approaches. The standards and samples preparation, as well as, derivatization and analysis were fully automated. For that purpose, a homemade chamber was built. The proposed method is based on liquid-liquid extraction of glycerol and simultaneous oxidation with periodate, generating formaldehyde that reacts with acetylacetone. A fluorescent product of 3,5-diacetyl-1,4-dihydrolutidine was obtained. The fluorescence signal was recorded at λ(ex) =417 nm and λ(em) = 514 nm. A linear response was observed from 0.10 to 5.00 mg L(-1) glycerol, variation coefficient 1.5%, sampling rate 14 h(-1) and detection limit 0.036 mg L(-1) glycerol. The procedure was successfully applied to the analysis of biodiesel samples, and the results agreed with the reference method (ASTM D6584-07) at 95% confidence level.
Sigma-Aldrich
Sodium (meta)periodate, BioUltra, ≥99.5% (RT)
Sigma-Aldrich
Sodium (meta)periodate, ≥99.0%
Sigma-Aldrich
Acetylacetone, JIS special grade, ≥99%
Sigma-Aldrich
Acetylacetone, produced by Wacker Chemie AG, Burghausen, Germany, ≥99.5% (GC)
Supelco
Acetylacetone, analytical standard
Sigma-Aldrich
Acetylacetone, ReagentPlus®, ≥99%
Sigma-Aldrich
Sodium periodate, ACS reagent, ≥99.8%