Merck
  • Resonant inelastic x-ray scattering of methyl chloride at the chlorine K edge.
Resonant inelastic x-ray scattering of methyl chloride at the chlorine K edge.
The Journal of chemical physics (2012-01-21)
L El Khoury, L Journel, R Guillemin, S Carniato, W C Stolte, T Marin, D W Lindle, M Simon
ABSTRACT
We present a combined experimental and theoretical study of isolated CH(3)Cl molecules using resonant inelastic x-ray scattering (RIXS). The high-resolution spectra allow extraction of information about nuclear dynamics in the core-excited molecule. Polarization-resolved RIXS spectra exhibit linear dichroism in the spin-orbit intensities, a result interpreted as due to chemical environment and singlet-triplet exchange in the molecular core levels. From analysis of the polarization-resolved data, Cl 2p(x, y) and 2p(z) electronic populations can be determined.
Sigma-Aldrich
Chloromethane, ≥99.5%
Supelco
Chloromethane solution, 200 μg/mL in methanol, analytical standard
Sigma-Aldrich
Chloromethane solution, 1.0 M in tert-butyl methyl ether, anhydrous
Sigma-Aldrich
Chloromethane solution, 1.0 M in diethyl ether