Merck
  • Validation of self-reported khat chewing amongst khat chewers: an exploratory study.
Validation of self-reported khat chewing amongst khat chewers: an exploratory study.
Journal of ethnopharmacology (2012-01-17)
Saba Kassim, Aamenah Hawash, Atholl Johnston, Ray Croucher
ABSTRACT
Khat chewing amongst the UK communities originating from Yemen and the East African coast is suggested to create dependency through its main stimulant components (cathinone, norephedrine and norpseudoephedrine) on the central nervous system. To validate self-reported khat chewing behaviours by measuring levels of cathinone, norephedrine and norpseudoephedrine in saliva and to explore their associations with self-reported khat chewing dependency. Face-to-face interviews were conducted amongst 30 male UK-resident khat chewers. Saliva samples were collected from each participant and high-performance liquid chromatography (HPLC) employed to extract and quantify the levels of the biomarkers. The mean (SD) for cathinone and the composite norephedrine and norpseudoephedrine levels were 33.93 (±39.20) and 29.28 (±26.32)μg/mL respectively. These biomarkers were significantly associated (p≤0.05) with khat chewing dependency. Validation of self-reported khat chewing is possible. Khat chewing dependency correlates significantly with biomarker levels in saliva. Replication is required.
Product Description
Supelco
(+)-Norpseudoephedrine hydrochloride solution, 100 μg/mL in methanol (as free base), ampule of 1 mL, certified reference material, Cerilliant®
Sigma-Aldrich
Cathine hydrochloride
Sigma-Aldrich
DL-Norephedrine hydrochloride, ≥99%
Supelco
(±)-Phenylpropanolamine hydrochloride solution, 1.0 mg/mL in methanol (as free base), ampule of 1 mL, certified reference material, Cerilliant®
Sigma-Aldrich
(1S,2R)-(+)-Norephedrine, 98%