Merck
  • Role and significance of extracellular polymeric substances on the property of aerobic granule.
Role and significance of extracellular polymeric substances on the property of aerobic granule.
Bioresource technology (2012-01-11)
Liang Zhu, Mei-le Lv, Xin Dai, Yan-wen Yu, Han-ying Qi, Xiang-yang Xu
ABSTRACT
Because of the important role of extracellular polymeric substances (EPS) in microbial aggregates, the relationship of the EPS and physico-chemical properties of aerobic granule was studied in two sequencing airlift bioreactors (SABR) using two kinds of substrates, acetate+glucose and 4-chloroaniline (4-ClA). Higher amounts of the exopolymeric protein (PN) and lower polysaccharides (PS) and DNA level were extracted from aerobic granules in the SABRs, and the variations in the PN and PN/PS of aerobic granules were higher (770% and 360%) in the SABR using 4-ClA as substrate than that in the other reactor. At the same time, suitable operating parameters (surface gas velocity at about 2.4 cm s(-1) and settling time at 5-10 min) favored EPS production and sludge granulation. Furthermore, the minimal settling velocity, hydrophobicity, surface charge increased in accordance with PN content increase, and a negative correlation between the sludge volume index (SVI) and PN content was observed.
Product Description
Supelco
p-Chloroaniline, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
4-Chloroaniline, 98%
Sigma-Aldrich
4-Chloroaniline, purified by sublimation, ≥99%
Supelco
4-Chloroaniline, PESTANAL®, analytical standard