Merck
  • Conjugated polymer nanoparticles for light-activated anticancer and antibacterial activity with imaging capability.
Conjugated polymer nanoparticles for light-activated anticancer and antibacterial activity with imaging capability.
Langmuir : the ACS journal of surfaces and colloids (2011-11-08)
Hui Chong, Chenyao Nie, Chunlei Zhu, Qiong Yang, Libing Liu, Fengting Lv, Shu Wang
ABSTRACT
A new water-soluble conjugated polymer containing fluorene and boron-dipyrromethene repeat units in the backbones (PBF) that exhibits red emission was synthesized and characterized. Cationic PBF forms uniform nanoparticles with negatively charged disodium salt 3,3'-dithiodipropionic acid (SDPA) in aqueous solution through electrostatic interactions. The nanoparticles display absorption maximum at 550 nm and emission maximum at 590 nm. Upon photoexcitation with white light (400-800 nm) with 90 and 45 mW·cm(-2) for bacteria and cancer cells killing respectively, PBF nanoparticles can sensitize the oxygen molecule to readily produce reactive oxygen species (ROS) for rapidly killing neighboring bacteria and cancer cells. Furthermore, PBF nanoparticles concurrently provide optical imaging capability. PBF nanoparticles are therefore a promising multifunctional material for treating cancers and bacteria infections, while concurrently providing optical monitoring capabilities.
Sigma-Aldrich
Tetramethylammonium chloride, reagent grade, ≥98%
Supelco
Tetramethylammonium chloride, suitable for ion pair chromatography, LiChropur, ≥99.0% (AT)
Sigma-Aldrich
Tetramethylammonium hydroxide solution, 10 wt. % in H2O
Sigma-Aldrich
Tetramethylammonium hydroxide solution, ACS reagent
Sigma-Aldrich
Tetramethylammonium fluoride, 97%
Supelco
Tetramethylammonium sulfate, suitable for ion pair chromatography, LiChropur, ≥99.0% (T)
Sigma-Aldrich
Tetramethylammonium hydroxide solution, 25 wt. % in methanol
Sigma-Aldrich
Tetramethylammonium hydroxide solution, 25 wt. % in H2O
Sigma-Aldrich
Tetramethylammonium chloride, BioUltra, for molecular biology, ≥99.0% (AT)
Sigma-Aldrich
Tetramethylammonium bromide, ACS reagent, ≥98.0%
Supelco
Tetramethylammonium bromide, suitable for ion pair chromatography, LiChropur, ≥99.0% (AT)
Sigma-Aldrich
Tetramethylammonium hydroxide pentahydrate, ≥95.0% (T)
Sigma-Aldrich
Tetramethylammonium iodide, 99%
Sigma-Aldrich
Tetramethylammonium bromide, 98%
Sigma-Aldrich
Tetramethylammonium hydroxide pentahydrate, ≥97%
Sigma-Aldrich
Tetramethylammonium chloride solution, for molecular biology