Merck
Fragrance material review on 2-phenoxyethanol.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association (2011-11-01)
J Scognamiglio, L Jones, C S Letizia, A M Api
ABSTRACT
A toxicologic and dermatologic review of 2-phenoxyethanol when used as a fragrance ingredient is presented. 2-Phenoxyethanol is a member of the fragrance structural group Aryl Alkyl Alcohols and is a primary alcohol. The AAAs are a structurally diverse class of fragrance ingredients that includes primary, secondary, and tertiary alkyl alcohols covalently bonded to an aryl (Ar) group, which may be either a substituted or unsubstituted benzene ring. The common structural element for the AAA fragrance ingredients is an alcohol group -C-(R1)(R2)OH and generically the AAA fragrances can be represented as an Ar-C-(R1)(R2)OH or Ar-Alkyl-C-(R1)(R2)OH group. This review contains a detailed summary of all available toxicology and dermatology papers that are related to this individual fragrance ingredient and is not intended as a stand-alone document. Available data for 2-phenoxyethanol were evaluated then summarized and includes physical properties, acute toxicity, skin irritation, mucous membrane (eye) irritation, skin sensitization, elicitation, phototoxicity, photoallergy, toxicokinetics, repeated dose, and reproductive toxicity data. A safety assessment of the entire Aryl Alkyl Alcohols will be published simultaneously with this document; please refer to Belsito et al. (2012) for an overall assessment of the safe use of this material and all Aryl Alkyl Alcohols in fragrances.
Product Description
USP
Phenoxyethanol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Ethylene glycol monophenyl ether, ≥90%
Phenoxyethanol, European Pharmacopoeia (EP) Reference Standard
Supelco
2-Phenoxyethanol, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
2-Phenoxyethanol, analytical standard
Sigma-Aldrich
2-Phenoxyethanol, ≥99%