Merck
Computational studies of lithium diisopropylamide deaggregation.
The Journal of organic chemistry (2011-09-06)
Alexander C Hoepker, David B Collum
ABSTRACT
Density functional theory computations [MP2/6-31G(d)//B3LYP/6-31G(d)] on the deaggregation of lithium diisopropylamide (LDA) dimer solvated by two tetrahydrofuran ligands to give the corresponding trisolvated monomer show eight structurally distinct minima. The barriers to exchange are comparable to those expected from experimental studies showing rate-limiting deaggregations. The role of conformational isomerism in deaggregation and the extent that deaggregation rates dictate LDA reactivity under synthetically important conditions are considered.
Sigma-Aldrich
Lithium diisopropylamide, 97%
Sigma-Aldrich
Diisopropylamine, SAJ first grade, ≥98.0%
Sigma-Aldrich
Lithium diisopropylamide solution, 2.0 M in THF/heptane/ethylbenzene
Sigma-Aldrich
Diisopropylamine, purified by redistillation, 99.95%
Supelco
Diisopropylamine, analytical standard
Sigma-Aldrich
Diisopropylamine, ≥99.5%
Sigma-Aldrich
Diisopropylamine, puriss. p.a., ≥99.0% (GC)