Merck
  • Bioremediation of 2-chlorophenol containing wastewater by aerobic granules-kinetics and toxicity.
Bioremediation of 2-chlorophenol containing wastewater by aerobic granules-kinetics and toxicity.
Journal of hazardous materials (2011-04-07)
Mohammad Zain Khan, Pijush Kanti Mondal, Suhail Sabir
ABSTRACT
2-Chlorophenol (2-CP) degrading aerobic granules were cultivated in a sequencing batch reactor (SBR) in presence of glucose. The organic loading rate (OLR) was increased from 6.9 to 9.7 kg COD m(-3)d(-1) (1150-1617 mg L(-1)COD per cycle) during the experiment. The alkalinity (1000 mg L(-1) as CaCO(3)) was maintained throughout the experiment. The specific cell growth rate was found to be 0.013 d(-1). A COD removal efficiency of 94% was achieved after steady state at 8h HRT (hydraulic retention time). FTIR, UV, GC, GC/MS studies confirmed that the biodegradation of 2-CP occurs via chlorocatechol (modified ortho-cleavage) pathway. Biodegradation kinetics followed the Haldane model with kinetic parameters: V(max)=840 mg2-CPgMLVSS(-1)d(-1), K(s)=24.61 mg L(-1), K(i)=315.02 mg L(-1). Abiotic losses of 2-CP due to volatilization and photo degradation by sunlight were less than 3% and the results of genotoxicity showed that the degradation products are eco-friendly.
Sigma-Aldrich
2-Chlorophenol, SAJ first grade, ≥99.0%
Sigma-Aldrich
2-Chlorophenol, 98%
Supelco
2-Chlorophenol, PESTANAL®, analytical standard
Sigma-Aldrich
2-Chlorophenol, ≥99%