Merck
  • Genome-wide expression analysis of human in vivo irritated epidermis: differential profiles induced by sodium lauryl sulfate and nonanoic acid.
Genome-wide expression analysis of human in vivo irritated epidermis: differential profiles induced by sodium lauryl sulfate and nonanoic acid.
The Journal of investigative dermatology (2010-04-30)
Anders Clemmensen, Klaus E Andersen, Ole Clemmensen, Qihua Tan, Thomas K Petersen, Torben A Kruse, Mads Thomassen
ABSTRACT
The pathogenesis of irritant contact dermatitis (ICD) is poorly understood, and genes participating in the epidermal response to chemical irritants are only partly known. It is commonly accepted that different irritants have different mechanisms of action in the development of ICD. To define the differential molecular events induced in the epidermis by different irritants, we collected sequential biopsies ((1/2), 4, and 24 hours after a single exposure and at day 11 after repeated exposure) from human volunteers exposed to either sodium lauryl sulfate (SLS) or nonanoic acid (NON). Gene expression analysis using high-density oligonucleotide microarrays (representing 47,000 transcripts) revealed essentially different pathway responses (1/2)hours after exposure: NON transiently induced the IL-6 pathway as well as a number of mitogen-activated signaling cascades including extracellular signal-regulated kinase and growth factor receptor signaling, whereas SLS transiently downregulated cellular energy metabolism pathways. Differential expression of the cyclooxygenase-2 and matrix metalloproteinase 3 transcripts was confirmed immunohistochemically. After cumulative exposure, 883 genes were differentially expressed, whereas we identified 23 suggested common biomarkers for ICD. In conclusion, we bring new insights into two hitherto less well-elucidated phases of skin irritancy: the very initial as well as the late phase after single and cumulative mild exposures, respectively.
Sigma-Aldrich
Nonanoic acid, ≥96%, FG
Sigma-Aldrich
Nonanoic acid, 96%
Supelco
Nonanoic acid, analytical standard
Sigma-Aldrich
Nonanoic acid, ≥97%