Merck
  • Non-extraction flow injection determination of cationic surfactants using eriochrome black-T.
Non-extraction flow injection determination of cationic surfactants using eriochrome black-T.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy (2009-05-05)
Ali A Ensafi, B Hemmateenejad, S Barzegar
ABSTRACT
A new, rapid, sensitive, non-extraction batch, and flow injection spectrophotometric method for the determination of cationic surfactants (CSs) such as cetyltrimethyl ammonium bromide (CTAB), tetra-n-butyl ammonium chloride (TBAC) and cetylpyridinium chloride (CPC) is proposed. The method is based on the interaction of cationic surfactants with eriochrome black-T to form an ion-association complex. This complex has strong absorbance at 708 nm. The effects of chemical parameters and FIA variables on the determination of cationic surfactants were studied in detail, especially for CTAB. Under optimum conditions, the two linear calibration ranges of the method are 3.0 x 10(-6) to 5.0 x 10(-3)mol L(-1) CTAB, CPB and DTAB for the batch spectrophotometric method and 2.0 x 10(-6) to 2.0 x 10(-4)mol L(-1) CTAB, CPB and TBC for the flow injection spectrophotometric method. The sample throughput was 35+/-5 samples h(-1) at room temperature. The relative standard deviations for 10 replicates of analysis of (2.0, 0.6 and 0.2)x10(-4)mol L(-1) CTAB were 1.2, 1.3, and 0.8%, respectively. In addition, the influence of potential interfering substances on the determination of cationic surfactants was studied. The proposed method is simple and rapid, using no toxic organic solvents. It was applied to the determination of trace CS in industrial wastewater with satisfactory results.
Sigma-Aldrich
Eriochrome® Black T solution
Sigma-Aldrich
Eriochrome® Black T, SAJ special grade
Sigma-Aldrich
Eriochrome® Black T, ACS reagent (indicator grade)