Merck
  • Microelectrode arrays and ceric ammonium nitrate: a simple strategy for developing new site-selective synthetic methods.
Microelectrode arrays and ceric ammonium nitrate: a simple strategy for developing new site-selective synthetic methods.
Journal of the American Chemical Society (2008-08-06)
David Kesselring, Karl Maurer, Kevin D Moeller
ABSTRACT
Conditions for a site-selective ceric ammonium nitrate oxidation have been developed. The reactions proceed nicely on both 1K- and 12K-microelectrode arrays. The procedure for developing the reactions was very simple and demonstrated that the same reagents used for a solution-phase reaction can be used for a related site-selective reaction on a microelectrode array.
Sigma-Aldrich
Ammonium cerium(IV) nitrate, JIS special grade, ≥95.0%
Sigma-Aldrich
Ammonium cerium(IV) nitrate, ≥99.99% trace metals basis
Sigma-Aldrich
Ammonium cerium(IV) nitrate, ACS reagent, ≥98.5%
Sigma-Aldrich
Ammonium cerium(IV) nitrate, puriss. p.a., ACS reagent, ≥98.5% (RT)
Sigma-Aldrich
Ammonium cerium(IV) nitrate, ≥98% (titration)