Merck
  • Study of the voltammetric behaviour of the ethalfluralin and methalpropalin and its determination in environmental matrices at hanging mercury drop electrode.
Study of the voltammetric behaviour of the ethalfluralin and methalpropalin and its determination in environmental matrices at hanging mercury drop electrode.
Environmental monitoring and assessment (2008-04-04)
T Thriveni, J Rajesh Kumar, Jin Young Lee, N Y Sreedhar
ABSTRACT
An electroanalytical method has been developed for the determination of the herbicides ethalfluralin[N-ethyl-N-(2-methyl-2-propenyl)-2,6-dinitro-4-(trifluoromethyl) bezenamine] and methalpropalin [N-(2-methyl-2-propenyl)-2, 6-dinitro-N-propyl-4 (trifluoromethyl) benzenamine] by differential pulse adsorptive stripping voltammetry (DP-AdSV) on a hanging mercury drop electrode (HMDE) with universal buffer as supporting electrolyte. The optimum adsorption conditions were found to be pH 6.0, an accumulation potential of -0.6 V (HMDE vs SCE), an accumulation time of 80 s. and scan rate 45 mVs(-1). Calibration curve is linear in the range 1.30 x 10(-9) to 1.32 x 10(-5) M of ethalfluralin and 1.13 x 10(-5) to 2.0 x 10(-8) M of methalpropalin with detection limits of 1.08 x 10(-9) and 1.87 x 10(-8) M, respectively. The relative SD and correlation coefficients were found to be 1.24%, 0.998 and 1.34%, 0.995, respectively for ten replicates. The method is applied to the determination of the ethalfluralin and methalpropalin in formulations and environmental matrices.
Sigma-Aldrich
α,α,α-Trifluorotoluene, anhydrous, ≥99%
Sigma-Aldrich
α,α,α-Trifluorotoluene, ≥99%
Sigma-Aldrich
α,α,α-Trifluorotoluene solution, NMR reference standard, 0.05% in benzene-d6 (99.6 atom % D), NMR tube size 3 mm × 8 in.
Supelco
Ethalfluralin, PESTANAL®, analytical standard
Sigma-Aldrich
α,α,α-Trifluorotoluene solution, NMR reference standard, 0.05% in benzene-d6 (99.6 atom % D), NMR tube size 10 mm × 8 in.
Sigma-Aldrich
α,α,α-Trifluorotoluene solution, NMR reference standard, 0.05% in benzene-d6 (99.6 atom % D)