Merck
  • Accurate molecular structures of chlorothiazide and hydrochlorothiazide by joint refinement against powder neutron and X-ray diffraction data.
Accurate molecular structures of chlorothiazide and hydrochlorothiazide by joint refinement against powder neutron and X-ray diffraction data.
Acta crystallographica. Section B, Structural science (2008-01-22)
Charlotte K Leech, Francesca P A Fabbiani, Kenneth Shankland, William I F David, Richard M Ibberson
ABSTRACT
The compounds chlorothiazide and hydrochlorothiazide (crystalline form II) have been studied in their fully hydrogenous forms by powder neutron diffraction on the GEM diffractometer. The results of joint Rietveld refinement of the structures against multi-bank neutron and single-bank X-ray powder data are reported and show that accurate and precise structural information can be obtained from polycrystalline molecular organic materials by this route.
Product Description
Supelco
Chlorothiazide, Pharmaceutical Secondary Standard; Certified Reference Material
Chlorothiazide, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Chlorothiazide, thiazide diuretic