Merck
  • Green and efficient synthesis of quinoxaline derivatives via ceric ammonium nitrate promoted and in situ aerobic oxidation of alpha-hydroxy ketones and alpha-keto oximes in aqueous media.
Green and efficient synthesis of quinoxaline derivatives via ceric ammonium nitrate promoted and in situ aerobic oxidation of alpha-hydroxy ketones and alpha-keto oximes in aqueous media.
Chemical & pharmaceutical bulletin (2008-01-08)
Ahmad Shaabani, Ali Maleki
ABSTRACT
The direct conversion of alpha-hydroxy ketones and alpha-keto oximes into quinoxaline derivatives in the presence of a catalytic amount of ceric ammonium nitrate via metal-catalyzed aerobic oxidation followed by in situ trapping with aromatic 1,2-diamines in water as a green and efficient reaction media, is reported.
Sigma-Aldrich
Ammonium cerium(IV) nitrate, JIS special grade, ≥95.0%
Sigma-Aldrich
Ammonium cerium(IV) nitrate, ≥99.99% trace metals basis
Sigma-Aldrich
Ammonium cerium(IV) nitrate, ≥98% (titration)
Sigma-Aldrich
Ammonium cerium(IV) nitrate, ACS reagent, ≥98.5%
Sigma-Aldrich
Ammonium cerium(IV) nitrate, puriss. p.a., ACS reagent, ≥98.5% (RT)