Merck
  • Production of natural methyl anthranilate by microbial N-demethylation of N-methyl methyl anthranilate by the topsoil-isolated bacterium Bacillus megaterium.
Production of natural methyl anthranilate by microbial N-demethylation of N-methyl methyl anthranilate by the topsoil-isolated bacterium Bacillus megaterium.
Journal of agricultural and food chemistry (2005-11-24)
Marcus Taupp, Dag Harmsen, Frank Heckel, Peter Schreier
ABSTRACT
Bacillus megaterium, isolated in a screening process from topsoil, was used for N-demethylation of natural N-methyl methyl anthranilate to produce natural methyl anthranilate. Maximal productivity of 70 mg/L/day was achieved under laboratory-scale conditions without further optimization. No byproducts were observed. Thus, production of "natural" methyl anthranilate using B. megaterium is a significant improvement over comparable already existing procedures.
Sigma-Aldrich
Methyl 2-aminobenzoate, ReagentPlus®, ≥99%
Sigma-Aldrich
Methyl anthranilate, ≥98%, FCC, FG