Merck
  • Economically pertinent continuous amide formation by direct lipase-catalyzed amidation with ammonia.
Economically pertinent continuous amide formation by direct lipase-catalyzed amidation with ammonia.
Biotechnology and bioengineering (2003-04-04)
Wouter F Slotema, Georgina Sandoval, David Guieysse, Adrie J J Straathof, Alain Marty
ABSTRACT
An economically pertinent process for the lipase-catalyzed synthesis of amides was developed. A continuous plug flow reactor was used. The model reaction was the production of oleamide, a lubricant and anti-slip agent, via direct Candida antarctica lipase B-catalyzed amidation of oleic acid with ammonia. Of all solvents tested, 2-methyl-2-butanol was found to respond optimally to the demands formulated in our specifications. A continuous conversion of oleic acid into oleamide of 85% was obtained. A productivity of 4.5 tons oleamide per kg of enzyme per year was calculated, indicating a contribution of enzyme to the product price of only 4%. The volumetric productivity, 100 g. L(-1). h(-1), is 4 to 100 times higher than in literature procedures. A simple crystallization procedure leads to 99% purity.
Sigma-Aldrich
2-Methyl-2-butanol, anhydrous, ≥99%
Supelco
2-Methyl-2-butanol, analytical standard
Sigma-Aldrich
2-Methyl-2-butanol, ReagentPlus®, 99%
Sigma-Aldrich
2-Methyl-2-butanol, ReagentPlus®, ≥99%