Merck
  • Sorption of nonpolar aromatic contaminants by chlorosilane surface modified natural minerals.
Sorption of nonpolar aromatic contaminants by chlorosilane surface modified natural minerals.
Environmental science & technology (2001-11-23)
P Huttenloch, K E Roehl, K Czurda
ABSTRACT
The efficacy of the surface modification of natural diatomite and zeolite material by chlorosilanes is demonstrated. Chlorosilanes used were trimethylchlorosilane (TMSCI), tert-butyldimethylchlorosilane (TBDMSCI), dimethyloctadecylchlorosilane (DMODSCI), and diphenyldichlorosilane (DPDSCI) possessing different headgroups and chemical properties. Silanol groups of the diatomite and zeolite were modified by chemical reaction with the chlorosilanes resulting in a stable covalent attachment of the organosilanes to the mineral surface. The alteration of surface properties of the modified material was proved by measurements of water adsorption capacity, total organic carbon (TOC) content, and thermoanalytical data. The surface modified material showed great stability even when exposed to extremes in ionic strength, pH, and to pure organic solvents. Sorption of toluene, o-xylene, and naphthalene from water was greatly enhanced by the surface modification compared to the untreated materials which showed no measurable sorption of these compounds. The enhanced sorption was dependent on the organic carbon content as well as on chemical characteristics of the chlorosilanes used. Batch sorption experiments showed that the phenyl headgroups of DPDSCI have the best affinity for aromatic compounds. Removal from an aqueous solution of 10 mg/L of naphthalene, o-xylene, and toluene was 71%, 60%, and 30% for surface modified diatomite and 51%, 30%, and 16% for modified clinoptilolite, respectively. Sorption data were well described by the Freundlich isotherm equation, which indicated physical adsorption onto the lipophilic surface rather than partitioning into the surface organic phase. The chlorosilane modified materials have an apparent potential for application in environmental technologies such as permeable reactive barriers (PRB) or wastewater treatment.
Sigma-Aldrich
tert-Butylchlorodimethylsilane solution, 50 wt. % in toluene
Sigma-Aldrich
Dichlorodiphenylsilane, 97%
Sigma-Aldrich
Dichlorodiphenylsilane, ≥95%
Sigma-Aldrich
tert-Butyldimethylsilyl chloride solution, 1.0 M in THF
Sigma-Aldrich
tert-Butyldimethylsilyl chloride, reagent grade, 97%