Merck
  • Determination of barbiturates in urine by micellar liquid chromatography and direct injection of sample.
Determination of barbiturates in urine by micellar liquid chromatography and direct injection of sample.
Journal of pharmaceutical and biomedical analysis (2000-03-07)
Y Martín-Biosca, S Sagrado, R M Villaneuva-Camañas, M J Medina-Hernández
ABSTRACT
A liquid chromatographic procedure for the determination of six barbiturates (barbital, diallyl barbituric acid, phenobarbital, butabarbital, amobarbital and pentobarbital) in urine samples is described. The proposed system uses a Spherisorb octadecyl-silane ODS-2 C18 analytical column and a guard column of similar characteristics. The UV detector was set at 240 nm. A study to select adequate composition of the micellar mobile phase for the separation of these compounds in urine samples is performed. Maximum resolution was achieved with a 0.07 M sodium dodecylsulphate-0.3% propanol at pH 7.4 eluent. Limits of detection at 240 nm were ranged between 0.13 microg ml(-1) for diallyl barbituric acid and 2.7 microg ml(-1) for amobarbital. The procedure allows for the determination of these compounds in 20 minutes, it does not require prior a sample preparation step and it can be very useful to the investigation of intoxication.
Product Description
Supelco
Butabarbital solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Sigma-Aldrich
Sodium 1-dodecanesulfonate, ReagentPlus®, ≥99%