Merck
  • Attenuation of massive cytokine response to the staphylococcal enterotoxin B superantigen by the innate immunomodulatory protein lactoferrin.
Attenuation of massive cytokine response to the staphylococcal enterotoxin B superantigen by the innate immunomodulatory protein lactoferrin.
Clinical and experimental immunology (2009-08-08)
J L Hayworth, K J Kasper, M Leon-Ponte, C A Herfst, D Yue, W C Brintnell, D M Mazzuca, D E Heinrichs, E Cairns, J Madrenas, D W Hoskin, J K McCormick, S M M Haeryfar
ABSTRACT
Staphylococcal enterotoxin B (SEB) is a pyrogenic exotoxin and a potent superantigen which causes massive T cell activation and cytokine secretion, leading to profound immunosuppression and morbidity. The inhibition of SEB-induced responses is thus considered a goal in the management of certain types of staphylococcal infections. Lactoferrin (LF) is a multi-functional glycoprotein with both bacteriostatic and bactericidal activities. In addition, LF is known to have potent immunomodulatory properties. Given the anti-microbial and anti-inflammatory properties of this protein, we hypothesized that LF can modulate T cell responses to SEB. Here, we report that bovine LF (bLF) was indeed able to attenuate SEB-induced proliferation, interleukin-2 production and CD25 expression by human leucocyte antigen (HLA)-DR4 transgenic mouse T cells. This inhibition was not due to bLF's iron-binding capacity, and could be mimicked by the bLF-derived peptide lactoferricin. Cytokine secretion by an engineered SEB-responsive human Jurkat T cell line and by peripheral blood mononuclear cells from healthy donors was also inhibited by bLF. These findings reveal a previously unrecognized property of LF in modulation of SEB-triggered immune activation and suggest a therapeutic potential for this naturally occurring protein during toxic shock syndrome.
Sigma-Aldrich
Lipopolysaccharides from Escherichia coli O55:B5, Ready Made solution, 1 mg/mL
Sigma-Aldrich
Lipopolysaccharides from Escherichia coli O55:B5, FITC conjugate
Sigma-Aldrich
Lipopolysaccharides from Escherichia coli O55:B5, purified by phenol extraction
Sigma-Aldrich
Lipopolysaccharides from Escherichia coli O55:B5, purified by gel-filtration chromatography
Sigma-Aldrich
Lipopolysaccharides from Escherichia coli O55:B5, purified by trichloroacetic acid extraction
Sigma-Aldrich
Lipopolysaccharides from Escherichia coli O55:B5, purified by ion-exchange chromatography, TLR ligand tested
Sigma-Aldrich
Lipopolysaccharides from Escherichia coli O55:B5, γ-irradiated, BioXtra, suitable for cell culture
Sigma-Aldrich
apo-Transferrin bovine, ≥98%
Sigma-Aldrich
apo-Transferrin bovine, BioReagent, suitable for cell culture, ≥98%