Merck
  • Efficient transfer of sialo-oligosaccharide onto proteins by combined use of a glycosynthase-like mutant of Mucor hiemalis endoglycosidase and synthetic sialo-complex-type sugar oxazoline.
Efficient transfer of sialo-oligosaccharide onto proteins by combined use of a glycosynthase-like mutant of Mucor hiemalis endoglycosidase and synthetic sialo-complex-type sugar oxazoline.
Biochimica et biophysica acta (2010-07-22)
Midori Umekawa, Takayuki Higashiyama, Yurie Koga, Tomonari Tanaka, Masato Noguchi, Atsushi Kobayashi, Shin-Ichiro Shoda, Wei Huang, Lai-Xi Wang, Hisashi Ashida, Kenji Yamamoto
ABSTRACT
An efficient method for synthesizing homogenous glycoproteins is essential for elucidating the structural and functional roles of glycans of glycoproteins. We have focused on the transglycosylation activity of endo-ß-N-acetylglucosaminidase from Mucor hiemalis (Endo-M) as a tool for glycoconjugate syntheses, since it can transfer en bloc the oligosaccharide of not only high-mannose type but also complex-type N-glycan onto various acceptors having an N-acetylglucosamine residue. However, there are two major bottlenecks for its practical application: the low yield of the transglycosylation product and the difficulty to obtain the activated sugar oxazoline substrate, especially the sialo-complex type one. We carried out the transglycosylation using a glycosynthase-like N175Q mutant of Endo-M, which was found to possess enhanced transglycosylation activity with sugar oxazoline as a donor substrate, in combination with an easy preparation of the sialo-complex-type sugar oxazoline from natural sialoglycopeptide in egg yolk. Endo-M-N175Q showed efficient transglycosylation toward sialo-complex-type sugar oxazoline onto bioactive peptides and bovine ribonuclease B, and each sialylated compound was obtained in significantly high yield. Highly efficient and simple chemo-enzymatic syntheses of various sialylated compounds were enabled, by a combination of a simple synthesis of sialo-complex-type sugar oxazoline and the Endo-M-N175Q catalyzed transglycosylation. Our method would be very useful for a practical synthesis of biologically important glycopeptides and glycoproteins.
Sigma-Aldrich
Endoglycosidase F1 from Elizabethkingia miricola, recombinant, expressed in E. coli, ≥16 U/mg, buffered aqueous solution
Sigma-Aldrich
Endoglycosidase H from Streptomyces plicatus, recombinant, expressed in E. coli, buffered aqueous solution
Sigma-Aldrich
Endoglycosidase F2 from Elizabethkingia miricola, recombinant, expressed in E. coli, 20 U/mg
Sigma-Aldrich
Endoglycosidase F3 from Elizabethkingia miricola, recombinant, expressed in E. coli, 30 U/mg