Merck
  • SAAG-4 is a novel mosquito salivary protein that programmes host CD4 T cells to express IL-4.
SAAG-4 is a novel mosquito salivary protein that programmes host CD4 T cells to express IL-4.
Parasite immunology (2009-06-06)
V D Boppana, S Thangamani, A J Adler, S K Wikel
ABSTRACT
Mosquitoes represent the most important vector for transmitting pathogens that cause human disease. Central to pathogen transmission is the ability to divert the host immune system away from Th1 and towards Th2 responsiveness. Identification of the mosquito factor(s) critical for programming Th2 responsiveness should therefore lead to strategies to neutralize their function and thus prevent disease transmission. In the current study, we used a TCR transgenic adoptive transfer system to screen gene products present in the saliva of the mosquito Aedes aegypti for their ability to programme CD4 T cells to express the signature Th2 cytokine IL-4. The clone SAAG-4 encodes a secreted protein with a predicted size of 20 kDa whose function has previously been uncharacterized. Notably, SAAG-4 reduced host CD4 T cell expression of the signature Th1 cytokine IFN-gamma while simultaneously increasing expression of IL-4. SAAG-4 is therefore the first identified mosquito factor that can programme Th2 effector CD4 T cell differentiation.
Sigma-Aldrich
GenElute HP Select Plasmid Gigaprep Kit, sufficient for 5 preparations
Sigma-Aldrich
Penicillin-Streptomycin, with 10,000 units penicillin and 10 mg streptomycin per mL in 0.9% NaCl, 0.1 μm filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Penicillin-Streptomycin, Solution stabilized, with 10,000 units penicillin and 10 mg streptomycin/mL, 0.1 μm filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Penicillin-Streptomycin, Solution Stabilized, with 5,000 units penicillin and 5mg streptomycin/mL, 0.1 μm filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Brefeldin A, ≥99% (HPLC and TLC), BioXtra, for molecular biology
Sigma-Aldrich
Brefeldin A, from Penicillium brefeldianum, Ready Made Solution, 10 mg/mL in DMSO
Sigma-Aldrich
Brefeldin A, from Penicillium brefeldianum, ≥99% (HPLC and TLC)