Merck
  • Multifunctional core-shell nanoparticles: discovery of previously invisible biomarkers.
Multifunctional core-shell nanoparticles: discovery of previously invisible biomarkers.
Journal of the American Chemical Society (2011-10-18)
Davide Tamburro, Claudia Fredolini, Virginia Espina, Temple A Douglas, Adarsh Ranganathan, Leopold Ilag, Weidong Zhou, Paul Russo, Benjamin H Espina, Giovanni Muto, Emanuel F Petricoin, Lance A Liotta, Alessandra Luchini
ABSTRACT
Many low-abundance biomarkers for early detection of cancer and other diseases are invisible to mass spectrometry because they exist in body fluids in very low concentrations, are masked by high-abundance proteins such as albumin and immunoglobulins, and are very labile. To overcome these barriers, we created porous, buoyant, core-shell hydrogel nanoparticles containing novel high affinity reactive chemical baits for protein and peptide harvesting, concentration, and preservation in body fluids. Poly(N-isopropylacrylamide-co-acrylic acid) nanoparticles were functionalized with amino-containing dyes via zero-length cross-linking amidation reactions. Nanoparticles functionalized in the core with 17 different (12 chemically novel) molecular baits showed preferential high affinities (K(D) < 10(-11) M) for specific low-abundance protein analytes. A poly(N-isopropylacrylamide-co-vinylsulfonic acid) shell was added to the core particles. This shell chemistry selectively prevented unwanted entry of all size peptides derived from albumin without hindering the penetration of non-albumin small proteins and peptides. Proteins and peptides entered the core to be captured with high affinity by baits immobilized in the core. Nanoparticles effectively protected interleukin-6 from enzymatic degradation in sweat and increased the effective detection sensitivity of human growth hormone in human urine using multiple reaction monitoring analysis. Used in whole blood as a one-step, in-solution preprocessing step, the nanoparticles greatly enriched the concentration of low-molecular weight proteins and peptides while excluding albumin and other proteins above 30 kDa; this achieved a 10,000-fold effective amplification of the analyte concentration, enabling mass spectrometry (MS) discovery of candidate biomarkers that were previously undetectable.
Sigma-Aldrich
Rhodamine 123, mitochondrial specific fluorescent dye
Sigma-Aldrich
Glucagon, ≥95% (HPLC), powder, synthetic
Sigma-Aldrich
Glucagon, synthetic, powder, suitable for cell culture
Sigma-Aldrich
Rhodamine 123, BioReagent, for fluorescence, ≥85% (HPLC)
Sigma-Aldrich
Potassium persulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Potassium persulfate, 99.99% trace metals basis
Sigma-Aldrich
Pararosaniline Base, crystalline
Supelco
Disperse Yellow 9, analytical standard
Sigma-Aldrich
Disperse Orange 3, Dye content 90 %
Sigma-Aldrich
Disperse Yellow 3, Dye content 30 %
Sigma-Aldrich
Acrylic acid, anhydrous, contains 200 ppm MEHQ as inhibitor, 99%
Sigma-Aldrich
Remazol Brilliant Blue R, anthraquinone dye
Supelco
Disperse Orange 3, analytical standard
Sigma-Aldrich
Disperse Blue 1, Dye content 30 %
Supelco
Disperse Yellow 3, analytical standard