Merck
  • LncRNA-SNHG5 mediates activation of hepatic stellate cells by regulating NF2 and Hippo pathway.
LncRNA-SNHG5 mediates activation of hepatic stellate cells by regulating NF2 and Hippo pathway.
Communications biology (2024-03-05)
Rongrong Zhang, Yating Zhan, Zhichao Lang, Yifei Li, Weizhi Zhang, Jianjian Zheng
ABSTRACT
Long noncoding RNA small nucleolar RNA host gene 5 (SNHG5) is an oncogene found in various human cancers. However, it is unclear what role SNHG5 plays in activating hepatic stellate cells (HSCs) and liver fibrosis. In this study, SNHG5 was found to be upregulated in activated HSCs in vitro and in primary HSCs isolated from fibrotic liver in vivo, and inhibition of SNHG5 suppressed HSC activation. Notably, Neurofibromin 2 (NF2), the main activator for Hippo signalling, was involved in the effects of SNHG5 on HSC activation. The interaction between SNHG5 and NF2 protein was further confirmed, and preventing the combination of the two could effectively block the effects of SNHG5 inhibition on EMT process and Hippo signaling. Additionally, higher SNHG5 was found in chronic hepatitis B patients and associated with the fibrosis stage. Altogether, we demonstrate that SNHG5 could serveĀ as an activated HSCs regulator via regulating NF2 and Hippo pathway.
Sigma-Aldrich
Anti-E-cadherin Antibody, clone DECMA-1, clone Decma-1, from rat
Sigma-Aldrich
Anti-BMP-7 Antibody, clone 2A10, clone 2A10, from mouse