Merck
  • Pretreatment with interleukin-15 attenuates inflammation and apoptosis by inhibiting NF-κB signaling in sepsis-induced myocardial dysfunction.
Pretreatment with interleukin-15 attenuates inflammation and apoptosis by inhibiting NF-κB signaling in sepsis-induced myocardial dysfunction.
European journal of histochemistry : EJH (2024-04-30)
Chaojie He, Yi Yu, Feifan Wang, Wudi Li, Hui Ni, Meixiang Xiang
ABSTRACT
Sepsis-induced myocardial dysfunction (SIMD) is associated with poor prognosis and increased mortality in patients with sepsis. Cytokines are important regulators of both the initiation and progression of sepsis. Interleukin-15 (IL-15), a pro-inflammatory cytokine, has been linked to protective effects against myocardial infarction and myocarditis. However, the role of IL-15 in SIMD remains unclear. We established a mouse model of SIMD via cecal ligation puncture (CLP) surgery and a cell model of myocardial injury via lipopolysaccharide (LPS) stimulation. IL-15 expression was prominently upregulated in septic hearts as well as cardiomyocytes challenged with LPS. IL-15 pretreatment attenuated cardiac inflammation and cell apoptosis and improved cardiac function in the CLP model. Similar cardioprotective effects of IL-15 pretreatment were observed in vitro. As expected, IL-15 knockdown had the opposite effect on LPS-stimulated cardiomyocytes. Mechanistically, we found that IL-15 pretreatment reduced the expression of the pro-apoptotic proteins cleaved caspase-3 and Bax and upregulated the anti-apoptotic protein Bcl-2. RNA sequencing and Western blotting further confirmed that IL-15 pretreatment suppressed the activation of nuclear factor kappa B (NF-κB) signaling in mice with sepsis. Besides, the addition of NF-κB inhibitor can significantly attenuate cardiomyocyte apoptosis compared to the control findings. Our results suggest that IL-15 pretreatment attenuated the cardiac inflammatory responses and reduced cardiomyocyte apoptosis by partially inhibiting NF-κB signaling in vivo and in vitro, thereby improving cardiac function in mice with sepsis. These findings highlight a promising therapeutic strategy for SIMD.
Sigma-Aldrich
Monoclonal Anti-BCL2 antibody produced in mouse, Prestige Antibodies® Powered by Atlas Antibodies, clone CL7935, purified immunoglobulin, buffered aqueous glycerol solution
Sigma-Aldrich
Anti-NF-κB p65 antibody produced in rabbit, affinity isolated antibody