Merck
  • Drosophila lifespan control by dietary restriction independent of insulin-like signaling.
Drosophila lifespan control by dietary restriction independent of insulin-like signaling.
Aging cell (2008-01-29)
Kyung-Jin Min, Rochele Yamamoto, Susanne Buch, Michael Pankratz, Marc Tatar
ABSTRACT
Reduced insulin/insulin-like growth factor (IGF) signaling may be a natural way for the reduction of dietary nutrients to extend lifespan. While evidence challenging this hypothesis is accumulating with Caenorhabditis elegans, for Drosophila melanogaster it is still thought that insulin/IGF and the mechanisms of dietary restriction (DR) might as yet function through overlapping mechanisms. Here, we aim to understand this potential overlap. We found that over-expression of dFOXO in head fat body extends lifespan and reduces steady-state mRNA abundance of insulin-like peptide-2 under conditions of high dietary yeast, but not when yeast is limiting. In contrast, conditions of DR that increase lifespan change only insulin-like peptide-5 (ilp5) mRNA abundance. Thus, reduction of ilp5 mRNA is associated with longevity extension by DR, while reduction of insulin-like peptide-2 is associated with the diet-dependent effects of FOXO over-expression upon lifespan. To assess whether reduction of ilp5 is required for DR to extend lifespan, we blocked its diet-dependent change with RNAi. Loss of the ilp5 dietary response did not diminish the capacity of DR to extend lifespan. Finally, we assessed the capacity of DR to extend lifespan in the absence of dFOXO, the insulin/IGF-responsive transcription factor. As with the knockdown of ilp5 diet responsiveness, DR was equally effective among genotypes with and without dFOXO. It is clear from many Drosophila studies that insulin/IGF mediates growth and metabolic responses to nutrition, but we now find no evidence that this endocrine system mediates the interaction between dietary yeast and longevity extension.
Sigma-Aldrich
Methyl 4-hydroxybenzoate, SAJ first grade, ≥98.0%
Sigma-Aldrich
Methyl 4-hydroxybenzoate, tested according to Ph. Eur.
Sigma-Aldrich
Methyl 4-hydroxybenzoate, ≥99%, FCC
Sigma-Aldrich
Methyl 4-hydroxybenzoate, analytical standard
Sigma-Aldrich
Methyl 4-hydroxybenzoate, ReagentPlus®, ≥99.0%, crystalline
Sigma-Aldrich
Methyl 4-hydroxybenzoate, BioXtra, ≥99.0% (titration)
Sigma-Aldrich
Methyl 4-hydroxybenzoate, BioReagent, suitable for insect cell culture
Sigma-Aldrich
Mifepristone, ≥98%