Merck
  • Synthesis and biological evaluation of 9-oxo-9H-indeno[1,2-b]pyrazine-2,3-dicarbonitrile analogues as potential inhibitors of deubiquitinating enzymes.
Synthesis and biological evaluation of 9-oxo-9H-indeno[1,2-b]pyrazine-2,3-dicarbonitrile analogues as potential inhibitors of deubiquitinating enzymes.
ChemMedChem (2010-02-27)
Matteo Colombo, Stefania Vallese, Ilaria Peretto, Xavier Jacq, Jean-Christophe Rain, Frédéric Colland, Philippe Guedat
ABSTRACT
High-throughput screening highlighted 9-oxo-9H-indeno[1,2-b]pyrazine-2,3-dicarbonitrile (1) as an active inhibitor of ubiquitin-specific proteases (USPs), a family of hydrolytic enzymes involved in the removal of ubiquitin from protein substrates. The chemical behavior of compound 1 was examined. Moreover, the synthesis and in vitro evaluation of new compounds, analogues of 1, led to the identification of potent and selective inhibitors of the deubiquitinating enzyme USP8.
Sigma-Aldrich
DUB-IN-1, ≥98% (HPLC)
Sigma-Aldrich
DUB-IN-2, ≥98% (HPLC)