Merck
  • Anti-hsa-miR-59 alleviates premature senescence associated with Hutchinson-Gilford progeria syndrome in mice.
Anti-hsa-miR-59 alleviates premature senescence associated with Hutchinson-Gilford progeria syndrome in mice.
The EMBO journal (2022-11-17)
Qianying Hu, Na Zhang, Tingting Sui, Guanlin Li, Zhiyao Wang, Mingyue Liu, Xiaojuan Zhu, Baiqu Huang, Jun Lu, Zhanjun Li, Yu Zhang
ABSTRACT
Hutchinson-Gilford progeria syndrome (HGPS) is a lethal premature aging disorder without an effective therapeutic regimen. Because of their targetability and influence on gene expression, microRNAs (miRNAs) are attractive therapeutic tools to treat diseases. Here we identified that hsa-miR-59 (miR-59) was markedly upregulated in HGPS patient cells and in multiple tissues of an HGPS mouse model (LmnaG609G/G609G ), which disturbed the interaction between RNAPII and TFIIH, resulting in abnormal expression of cell cycle genes by targeting high-mobility group A family HMGA1 and HMGA2. Functional inhibition of miR-59 alleviated the cellular senescence phenotype of HGPS cells. Treatment with AAV9-mediated anti-miR-59 reduced fibrosis in the quadriceps muscle, heart, and aorta, suppressed epidermal thinning and dermal fat loss, and yielded a 25.5% increase in longevity of LmnaG609G/G609G mice. These results identify a new strategy for the treatment of HGPS and provide insight into the etiology of HGPS disease.
Sigma-Aldrich
Anti-β-Actin antibody, Mouse monoclonal, clone AC-15, purified from hybridoma cell culture
Sigma-Aldrich
Monoclonal Anti-HA tag antibody produced in mouse, clone GT423, affinity isolated antibody