Merck
  • Mitochondrial fission links ECM mechanotransduction to metabolic redox homeostasis and metastatic chemotherapy resistance.
Mitochondrial fission links ECM mechanotransduction to metabolic redox homeostasis and metastatic chemotherapy resistance.
Nature cell biology (2022-02-16)
Patrizia Romani, Nunzia Nirchio, Mattia Arboit, Vito Barbieri, Anna Tosi, Federica Michielin, Soichi Shibuya, Thomas Benoist, Danchen Wu, Charles Colin Thomas Hindmarch, Monica Giomo, Anna Urciuolo, Flavia Giamogante, Antonella Roveri, Probir Chakravarty, Marco Montagner, Tito Calì, Nicola Elvassore, Stephen L Archer, Paolo De Coppi, Antonio Rosato, Graziano Martello, Sirio Dupont
ABSTRACT
Metastatic breast cancer cells disseminate to organs with a soft microenvironment. Whether and how the mechanical properties of the local tissue influence their response to treatment remains unclear. Here we found that a soft extracellular matrix empowers redox homeostasis. Cells cultured on a soft extracellular matrix display increased peri-mitochondrial F-actin, promoted by Spire1C and Arp2/3 nucleation factors, and increased DRP1- and MIEF1/2-dependent mitochondrial fission. Changes in mitochondrial dynamics lead to increased production of mitochondrial reactive oxygen species and activate the NRF2 antioxidant transcriptional response, including increased cystine uptake and glutathione metabolism. This retrograde response endows cells with resistance to oxidative stress and reactive oxygen species-dependent chemotherapy drugs. This is relevant in a mouse model of metastatic breast cancer cells dormant in the lung soft tissue, where inhibition of DRP1 and NRF2 restored cisplatin sensitivity and prevented disseminated cancer-cell awakening. We propose that targeting this mitochondrial dynamics- and redox-based mechanotransduction pathway could open avenues to prevent metastatic relapse.
Sigma-Aldrich
(−)-Blebbistatin, solid, synthetic
Sigma-Aldrich
Wortmannin, from Penicillium funiculosum, ≥98% (HPLC and TLC)
Sigma-Aldrich
Mdivi-1, ≥98% (HPLC), powder
Sigma-Aldrich
1S,3R-RSL 3, ≥98% (HPLC)
Sigma-Aldrich
N-Acetyl-L-cysteine, Sigma Grade, ≥99% (TLC), powder
Sigma-Aldrich
ML-7, powder
Sigma-Aldrich
MitoTEMPO, ≥98% (HPLC)
Sigma-Aldrich
KI-696, ≥98% (HPLC)