Merck
  • Sensory nerve niche regulates mesenchymal stem cell homeostasis via FGF/mTOR/autophagy axis.
Sensory nerve niche regulates mesenchymal stem cell homeostasis via FGF/mTOR/autophagy axis.
Nature communications (2023-01-21)
Fei Pei, Li Ma, Junjun Jing, Jifan Feng, Yuan Yuan, Tingwei Guo, Xia Han, Thach-Vu Ho, Jie Lei, Jinzhi He, Mingyi Zhang, Jian-Fu Chen, Yang Chai
ABSTRACT
Mesenchymal stem cells (MSCs) reside in microenvironments, referred to as niches, which provide structural support and molecular signals. Sensory nerves are niche components in the homeostasis of tissues such as skin, bone marrow and hematopoietic system. However, how the sensory nerve affects the behavior of MSCs remains largely unknown. Here we show that the sensory nerve is vital for mesenchymal tissue homeostasis and maintenance of MSCs in the continuously growing adult mouse incisor. Loss of sensory innervation leads to mesenchymal disorder and a decrease in MSCs. Mechanistically, FGF1 from the sensory nerve directly acts on MSCs by binding to FGFR1 and activates the mTOR/autophagy axis to sustain MSCs. Modulation of mTOR/autophagy restores the MSCs and rescues the mesenchymal tissue disorder of Fgfr1 mutant mice. Collectively, our study provides insights into the role of sensory nerves in the regulation of MSC homeostasis and the mechanism governing it.
Sigma-Aldrich
Tamoxifen, ≥99%
Sigma-Aldrich
Anti-FGFR1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Anti-Tyrosine Hydroxylase Antibody, Chemicon®, from rabbit